Skip to Content
Merck
All Photos(2)

Key Documents

302120

Sigma-Aldrich

n-Butyllithium solution

2.0 M in cyclohexane

Synonym(s):

n-BuLi, Butyl lithium, Butyllithium solution, Lithium-1-butanide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3Li
CAS Number:
Molecular Weight:
64.06
Beilstein:
1209227
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

form

liquid

Quality Level

concentration

2.0 M in cyclohexane

bp

80 °C

density

0.775 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

[Li]CCCC

InChI

1S/C4H9.Li/c1-3-4-2;/h1,3-4H2,2H3;

InChI key

MZRVEZGGRBJDDB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

n-Butyllithium (n-BuLi) is an organolithium reagent widely used as a strong base (superbase) in organic synthesis for the preparation of various chemical intermediates. It is also used as a reagent for lithium-halogen exchange and lithium-metal transmetalation reactions. n-BuLi is capable of lithiating carbon acids.

Application

n-Butyllithium (2.0 M in cyclohexane) can be used as:      
  • A polymerization initiator to synthesize polystyrenes by anionic polymerization of styrene.      
  • A strong base in the diastereoselective alkylation reactions.
  • A reagent to synthesize 2-benzoylpyrroles by reacting benzaldehydes with di(1H-pyrrol-1-yl)zirconium(IV) chloride complex.

Packaging

The 25 mL Sure/Seal bottle is recommended as a single-use bottle. Repeated punctures will likely result in decreased performance of product.

Legal Information

Sure/Seal is a trademark of Sigma-Aldrich Co. LLC

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3 - Water-react 1

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 2

Flash Point(F)

-0.4 °F - closed cup

Flash Point(C)

-18 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

High-field proton NMR study of the aggregation and complexation of n-butyllithium in tetrahydrofuran.
McGarrity JF and Ogle CA.
Journal of the American Chemical Society, 107(7), 1805-1810 (1985)
Comparison of the synthetic utility of n-butyllithium and lithium diisopropylamide in the metalations of N, N-dialkyltoluamides.
Ludt RE, et al
The Journal of Organic Chemistry, 38(9), 1668-1674 (1973)
Lithium intercalation via n-butyllithium of the layered transition metal dichalcogenides.
Dines MB.
Materials Research Bulletin, 10(4), 287-291 (1975)
Arene-metal complexes. 13. Reaction of substituted (benzene) tricarbonylchromium complexes with n-butyllithium.
Card RJ and Trahanovsky WS.
The Journal of Organic Chemistry, 45(13), 2560-2566 (1980)
Deyu Li et al.
The Journal of organic chemistry, 73(6), 2373-2381 (2008-02-26)
A system consisting of a chiral lithium amide and n-BuLi in tol-d(8) solution was investigated with (1)H and (13)C INEPT DOSY, (6)Li and (15)N NMR, and other 2D NMR techniques. A mixed 2:1 trimeric complex was identified as the major

Articles

Transformative reagents enable selective conversions within molecules containing sensitive functionalities under mild reactions.

Transformative reagents enable selective conversions within molecules containing sensitive functionalities under mild reactions.

Transformative reagents enable selective conversions within molecules containing sensitive functionalities under mild reactions.

Transformative reagents enable selective conversions within molecules containing sensitive functionalities under mild reactions.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service