Skip to Content
Merck
All Photos(2)

Key Documents

702722

Sigma-Aldrich

Potassium bis(trimethylsilyl)amide solution

1 M in THF

Synonym(s):

KHMDS, Hexamethyldisilazane potassium salt solution, Potassium hexamethyldisilazide solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)3Si]2NK
CAS Number:
Molecular Weight:
199.48
Beilstein:
4006754
MDL number:
UNSPSC Code:
12352111
PubChem Substance ID:
NACRES:
NA.22

vapor density

3.2 (vs air)

Quality Level

form

liquid

concentration

1 M in THF

density

0.915 g/mL at 25 °C

SMILES string

C[Si](C)(C)N([K])[Si](C)(C)C

InChI

1S/C6H18NSi2.K/c1-8(2,3)7-9(4,5)6;/h1-6H3;/q-1;+1

InChI key

IUBQJLUDMLPAGT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Potassium bis(trimethylsilyl)amide solution (1 M in THF) has been used as an anionic initiator in the polymerization of polyethylene oxide(PEO).

Packaging

The 25 mL Sure/Seal bottle is recommended as a single-use bottle. Repeated punctures will likely result in decreased performance of product.

Legal Information

Sure/Seal is a trademark of Sigma-Aldrich Co. LLC

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-2.0 °F

Flash Point(C)

-18.9 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Highly stable multi-anchored magnetic nanoparticles for optical imaging within biofilms
Stone R C
Journal of Colloid and Interface Science, 459, 175-182 (2015)
Adrienne C Greene et al.
Biomaterials, 32(34), 8860-8869 (2011-09-03)
Chemical and topographical cues can be used to guide dissociated neurons into user-defined network geometries on artificial substrates, yet control of neuron polarity (differentiation into axons and dendrites) remains an elusive goal. We developed a dual guidance cue strategy for
André M Bienfait et al.
Dalton transactions (Cambridge, England : 2003), 43(46), 17324-17332 (2014-10-17)
Trivalent Eu[N(SiHMe2)2]3(THF)2 can easily be synthesized by applying a routine salt metathesis protocol (EuCl3(THF)2 and 3 equiv. of Li[N(SiHMe2)2] in n-hexane) which crystallizes isotypically to its analogues of the rare-earth metal series (space group P21/c). Transsilylamination of Eu[N(SiMe3)2]2(THF)2 with a
Nur Hazlin Hazrin-Chong et al.
Journal of microbiological methods, 90(2), 96-99 (2012-05-09)
The use of hexamethyldisilazane (HMDS) as a drying agent was investigated in the specimen preparation for scanning electron microscopy (SEM) imaging of bacterial surface colonization on sub-bituminous coal. The ability of microbes to biofragment, ferment and generate methane from coal
Elsa Vennat et al.
Dental materials : official publication of the Academy of Dental Materials, 25(6), 729-735 (2009-01-29)
The objectives of this study were to assess demineralized dentin porosity and quantify the different porous features distribution within the material using mercury intrusion porosimetry (MIP) technique. We compared hexamethyldisilazane (HMDS) drying and lyophilization (LYO) (freeze-drying) in sample preparation. Fifty-six

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service