Skip to Content
Merck
All Photos(1)

Documents

20159

Sigma-Aldrich

n-Butyllithium solution

2.7 M in heptane

Synonym(s):

n-BuLi, Butyl lithium, Butyllithium solution, Lithium-1-butanide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3Li
CAS Number:
Molecular Weight:
64.06
Beilstein:
1209227
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

form

liquid

shelf life

limited shelf life

concentration

2.7 M in heptane

color

yellow

density

0.71 g/mL at 20 °C

storage temp.

2-8°C

SMILES string

[Li]CCCC

InChI

1S/C4H9.Li/c1-3-4-2;/h1,3-4H2,2H3;

InChI key

MZRVEZGGRBJDDB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

n-Butyllithium is a strong nucleophile in the synthetic organic chemistry. It is also used as an initiator in the polymerization process.

Caution

a precipitate of lithium hydride can form on longer storage

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3 - Water-react 1

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Tatiana Guzzo et al.
Biomolecules, 10(11) (2020-10-30)
The stability and the degradation of polymers in physiological conditions are very important issues in biomedical applications. The copolymer of hyaluronic acid and poly-D,L-lactic acid (made available in a product called DAC®) produces a hydrogel which retains the hydrophobic character
Methylsulfinyl carbanion (CH3-SO-CH2-). Formation and applications to organic synthesis
Corey EJ and Chaykovsky M
Journal of the American Chemical Society, 87(6), 1345-1353 (1965)
Some trends observed in [60] fullerene polymerization activated by butyllithium isomers
Atovmyan EG, et al.
Russian Chemical Bulletin, 61(11), 2178-2179 (2012)
Petra Rönnholm et al.
Organic letters, 9(19), 3781-3783 (2007-08-19)
A robust and scaleable route to chiral 1-isopropylamino-2-(diphenylphosphino)ethanes is described via the ring-opening of chiral, cyclic sulfamidates with potassium diphenylphosphide (KPPh(2)). The novel protocol offers a robust access to gram quantities of chiral amino phosphinoethanes in high yields. The Li-amides
Deyu Li et al.
The Journal of organic chemistry, 73(6), 2373-2381 (2008-02-26)
A system consisting of a chiral lithium amide and n-BuLi in tol-d(8) solution was investigated with (1)H and (13)C INEPT DOSY, (6)Li and (15)N NMR, and other 2D NMR techniques. A mixed 2:1 trimeric complex was identified as the major

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service