Skip to Content
Merck
All Photos(1)

Key Documents

309966

Sigma-Aldrich

Diethyl ether

≥99.9%, suitable for HPLC, inhibitor-free

Synonym(s):

Et2O, Ethoxyethane, Ether, Ethyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2)2O
CAS Number:
Molecular Weight:
74.12
Beilstein:
1696894
EC Number:
MDL number:
UNSPSC Code:
12190000
PubChem Substance ID:
NACRES:
NA.07
grade:
HPLC grade
technique(s):
HPLC: suitable
application(s):
food and beverages
bp:
34.6 °C (lit.)
vapor pressure:
28.69 psi ( 55 °C)
8.59 psi ( 20 °C)

Product Name

Diethyl ether, suitable for HPLC, ≥99.9%, inhibitor-free

grade

HPLC grade

Quality Level

vapor density

2.6 (vs air)

vapor pressure

28.69 psi ( 55 °C)
8.59 psi ( 20 °C)

Assay

≥99.9%

form

liquid

autoignition temp.

320 °F

purified by

glass distillation

expl. lim.

36.5 %

technique(s)

HPLC: suitable

impurities

≤0.03% water
≤1 ppm peroxides (as H2O2)

evapn. residue

<0.0003%

refractive index

n20/D 1.3530 (lit.)

bp

34.6 °C (lit.)

mp

−116 °C (lit.)

density

0.706 g/mL at 25 °C (lit.)

λ

H2O reference

UV absorption

λ: 218 nm Amax: 1.0
λ: 250 nm Amax: 0.20
λ: 275 nm Amax: 0.04
λ: 300-400 nm Amax: 0.01

application(s)

food and beverages

SMILES string

CCOCC

InChI

1S/C4H10O/c1-3-5-4-2/h3-4H2,1-2H3

InChI key

RTZKZFJDLAIYFH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Luminescence of In(III)Cl-etioporphyrin-I.: This study investigates the luminescence properties of In(III)Cl-etioporphyrin-I, employing diethyl ether in the synthesis process to enhance solubility and reaction outcomes (Koptyaev et al., 2023).


  • An efficient synthesis of phosphonated cyclopentenones by NaN(3)-catalyzed three-component reaction between trialkyl phosphites, ethyl arylmethylidenecyanoacetates and dialkyl acetylenedicarboxylates.: Highlights a novel synthesis technique using diethyl ether as a solvent to facilitate the reaction process, crucial for the production of phosphonated cyclopentenones (Ranjbar Derranji and Anary-Abbasinejad, 2024).


  • Antioxidant and Anti-Inflammatory Activities of Phenolic Acid-Rich Extract from Hairy Roots of Dracocephalum moldavica.: Discusses the extraction of phenolic acids using diethyl ether from Dracocephalum moldavica, examining their antioxidant and anti-inflammatory properties (Weremczuk-Jeżyna et al., 2023).

  • [An improved extraction and nonradioactive thin-layer chromatography detection method of mycolic acid].: Describes an improved method for the extraction and analysis of mycolic acid using diethyl ether, enhancing the detection efficiency in biological samples (Xu et al., 2023).


  • A Modified (1)H-NMR Quantification Method of Ephedrine Alkaloids in Ephedrae Herba Samples.: Presents a modified NMR quantification method where diethyl ether plays a critical role in the extraction and preparation of samples for precise measurement of ephedrine alkaloids (Li et al., 2023).


Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 1 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-40.0 °F - closed cup

Flash Point(C)

-40 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Lana S Barkawi et al.
Nature protocols, 5(10), 1609-1618 (2010-10-05)
Auxin measurements in plants are critical to understanding both auxin signaling and metabolic homeostasis. The most abundant natural auxin is indole-3-acetic acid (IAA). This protocol is for the precise, high-throughput determination of free IAA in plant tissue by isotope dilution
Diethyl ether (DEE) as a renewable diesel fuel.
Bailey B, et al.
No. 972978. SAE technical paper (1997)
Eagleson M.
Concise Encyclopedia Chemistry, 471-471 (1994)
Oxidation of secondary alcohols in diethyl ether with aqueous chromic acid. Convenient procedure for the preparation of ketones in high epimeric purity.
Brown HC, et al.
The Journal of Organic Chemistry, 36(3), 387-390 (1971)
Tatsuya Yoshino et al.
Organic letters, 14(16), 4290-4292 (2012-08-09)
A highly efficient total synthesis of the 11-membered cyclic aspercyclides A (1) and B (2) has been achieved by chemo- and regioselective intramolecular oxidative C-O bond formation from differently substituted diphenols.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service