Merck
  • Design and development of a novel vaccine for protection against Lyme borreliosis.
Design and development of a novel vaccine for protection against Lyme borreliosis.
PloS one (2014-11-20)
Pär Comstedt, Markus Hanner, Wolfgang Schüler, Andreas Meinke, Urban Lundberg
ABSTRACT
There is currently no Lyme borreliosis vaccine available for humans, although it has been shown that the disease can be prevented by immunization with an OspA-based vaccine (LYMErix). Outer surface protein A (OspA) is one of the dominant antigens expressed by the spirochetes when present in a tick. The Borrelia species causing Lyme borreliosis in Europe express different OspA serotypes on their surface, B. burgdorferi (serotype 1), B. afzelii (serotype 2), B. garinii (serotypes, 3, 5 and 6) and B. bavariensis (serotype 4), while only B. burgdorferi is present in the US. In order to target all these pathogenic Borrelia species, we have designed a multivalent OspA-based vaccine. The vaccine includes three proteins, each containing the C-terminal half of two OspA serotypes linked to form a heterodimer. In order to stabilize the C-terminal fragment and thus preserve important structural epitopes at physiological temperature, disulfide bonds were introduced. The immunogenicity was increased by introduction of a lipidation signal which ensures the addition of an N-terminal lipid moiety. Three immunizations with 3.0 µg adjuvanted vaccine protected mice from a challenge with spirochetes expressing either OspA serotype 1, 2 or 5. Mice were protected against both challenge with infected ticks and in vitro grown spirochetes. Immunological analyses (ELISA, surface binding and growth inhibition) indicated that the vaccine can provide protection against the majority of Borrelia species pathogenic for humans. This article presents the approach which allows for the generation of a hexavalent vaccine that can potentially protect against a broad range of globally distributed Borrelia species causing Lyme borreliosis.
Product Description
Imidazole, European Pharmacopoeia (EP) Reference Standard
USP
Imidazole, United States Pharmacopeia (USP) Reference Standard
Supelco
Imidazole, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
EZBlue Gel Staining Reagent
Sigma-Aldrich
Hydroxyapatite, puriss., meets analytical specification of Ph. Eur., BP, FCC, E341, ≥90% (calculated on glowed substance)
Sigma-Aldrich
Calcium phosphate tribasic, BioReagent, suitable for plant cell culture, powder
Sigma-Aldrich
Calcium phosphate tribasic, 34.0-40.0% Ca basis
Sigma-Aldrich
Imidazole, ReagentPlus®, 99%, Redi-Dri, free-flowing
Sigma-Aldrich
Imidazole, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Imidazole, Molecular Biology, ≥99% (titration)
Sigma-Aldrich
Imidazole, Molecular Biology, ≥99% (titration), free-flowing, Redi-Dri
Sigma-Aldrich
Hydroxyapatite, nanopowder, <200 nm particle size (BET), contains 5 wt. % silica as dopant, synthetic
Sigma-Aldrich
Imidazole, ≥99% (titration), crystalline
Sigma-Aldrich
Imidazole, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Hydroxyapatite, nanoparticles, dispersion, 10 wt. % in H2O, <200 nm particle size (BET)
Sigma-Aldrich
Hydroxyapatite, synthetic, 99.8% trace metals basis (excludes Mg)
Sigma-Aldrich
Imidazole, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Imidazole, ACS reagent, ≥99% (titration)
Sigma-Aldrich
Imidazole, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Hydroxyapatite, purum p.a., ≥90% (as Ca3(PO4)2, KT)
Sigma-Aldrich
Hydroxyapatite, powder, synthetic
Sigma-Aldrich
Imidazole, ReagentPlus®, 99%
Sigma-Aldrich
Hydroxyapatite, nanopowder, <200 nm particle size (BET), ≥97%, synthetic
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
TWEEN® 20, Molecular Biology, viscous liquid
Sigma-Aldrich
Triton X-114
Sigma-Aldrich
Sephadex® G-25, BioReagent, DNA grade, Molecular Biology, Superfine
Ondansetron impurity E, European Pharmacopoeia (EP) Reference Standard
SAFC
Isopropyl β-D-1-thiogalactopyranoside
Custom Peptides, Product Overview, Design, and Ordering