Skip to Content
Merck

Skip To

1336500

USP

Imidazole

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

1,3-Diaza-2,4-cyclopentadiene, Glyoxaline

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
C3H4N2
CAS Number:
Molecular Weight:
68.08
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
103853
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

vapor pressure

<1 mmHg ( 20 °C)

API family

imidazole

manufacturer/tradename

USP

pKa (25 °C)

6.95

bp

256 °C (lit.)

mp

88-91 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

c1c[nH]cn1

InChI

1S/C3H4N2/c1-2-5-3-4-1/h1-3H,(H,4,5)

InChI key

RAXXELZNTBOGNW-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Excellent for buffers in the range of pH 6.2-7.8
Imidazole USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Clotrimazole
  • Clotrimazole Topical Solution
  • Perindopril Erbumine
  • Moexipril Hydrochloride
  • Clotrimazole Vaginal Inserts
  • Sildenafil Citrate
  • Clotrimazole Lozenges

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
I0086000PHR1180I5513
manufacturer/tradename

USP

manufacturer/tradename

EDQM

manufacturer/tradename

-

manufacturer/tradename

-

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

certified reference material, pharmaceutical secondary standard

grade

Molecular Biology

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

diagnostic assay manufacturing
genomic analysis
life science and biopharma
microbiology

format

neat

format

neat

format

neat

format

-

bp

256 °C (lit.)

bp

256 °C (lit.)

bp

256 °C (lit.)

bp

256 °C (lit.)

vapor pressure

<1 mmHg ( 20 °C)

vapor pressure

<1 mmHg ( 20 °C)

vapor pressure

<1 mmHg ( 20 °C)

vapor pressure

<1 mmHg ( 20 °C)


Still not finding the right product?

Explore all of our products under Imidazole


signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Repr. 1B - Skin Corr. 1C

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

flash_point_f

293.0 °F - closed cup

flash_point_c

145 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters