Skip to Content
Merck
All Photos(1)

Key Documents

702153

Sigma-Aldrich

Hydroxyapatite

nanoparticles, dispersion, 10 wt. % in H2O, <200 nm particle size (BET)

Synonym(s):

Calcium hydroxide phosphate, Calcium hydroxylapatite, Calcium hydroxyphosphate, Nano hydroxyapatatite, Pentacalcium hydroxide triphosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[Ca5(OH)(PO4)3]x
CAS Number:
Molecular Weight:
502.31
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
dispersion
nanoparticles

grade

for analytical purposes

form

dispersion
nanoparticles

contains

≤0.025 wt. % dispersant (non-metal based)

concentration

10 wt. % in H2O

surface area

14.3 m2/g , typical

particle size

<200 nm (BET)

pH

4-6

density

1.038 g/mL at 25 °C

SMILES string

[Ca++].[Ca++].[Ca++].[Ca++].O[Ca+].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O

InChI

1S/5Ca.3H3O4P.H2O/c;;;;;3*1-5(2,3)4;/h;;;;;3*(H3,1,2,3,4);1H2/q5*+2;;;;/p-10

InChI key

XYJRXVWERLGGKC-UHFFFAOYSA-D

Looking for similar products? Visit Product Comparison Guide

General description

Hydroxyapatite(HAP) is an osteoconductive bioceramic material which has uniqueability to form direct bonds with living tissues. It can be easily prepared byvarious techniques such as sol-gel, hydrothermal, multiple emulsion, biomimeticdeposition, and electrodeposition.

Application

Hydroxyapatite can be used as:
  • An implant material for bone substitute due to its chemical similarity to the inorganic component of bone and osteo inductive properties.
  • A vehicle for gene and drug delivery because of its encapsulation capacity and affinity to several drugs and proteins.
  • A pH responsive particulate emulsifier.

Features and Benefits

  • Biocompatible
  • Osteoconductive
  • Stable under physiological conditions
  • High mechanical strength

Legal Information

Product of Engi-Mat Co.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Martin P Reichhardt et al.
Life science alliance, 3(4) (2020-02-27)
The scavenger receptor cysteine-rich (SRCR) family of proteins comprises more than 20 membrane-associated and secreted molecules. Characterised by the presence of one or more copies of the ∼110 amino-acid SRCR domain, this class of proteins have widespread functions as antimicrobial
W Zhang et al.
Journal of dental research, 92(12), 1136-1141 (2013-10-08)
Stem cell-based bone tissue engineering has been recognized as a new strategy for maxillary sinus floor elevation. More rapid bone formation may enhance this technique when simultaneous dental implant placement is desired. Adipose tissue-derived stem cells (ADSCs) and bone marrow
Satyavrata Samavedi et al.
Acta biomaterialia, 9(9), 8037-8045 (2013-06-25)
Calcium phosphate ceramics (CPCs) have been widely used as biomaterials for the regeneration of bone tissue because of their ability to induce osteoblastic differentiation in progenitor cells. Despite the progress made towards fabricating CPCs possessing a range of surface features
Liam M Grover et al.
Biomaterials, 34(28), 6631-6637 (2013-06-12)
Pyrophosphate ions are both inhibitors of HA formation and substrates for phosphatase enzymes. Unlike polyphosphates their hydrolysis results simultaneously in the complete loss of mineral formation inhibition and a localised elevation in orthophosphate ion concentration. Despite recent advances in our
Xi Chen et al.
The Journal of biological chemistry, 289(16), 11162-11174 (2014-03-13)
Calcium phosphate precipitates (CPPs) form complexes with DNA, which enter cells via endocytosis. Under this condition CPPs induce autophagy via the canonic autophagy machinery. Here we showed that CPP-induced autophagy was also dependent on endocytosis as the process was significantly

Articles

Nanoparticles can be used for applications such as growing cells for tissue engineering, nanocomposites used in orthopedics, and miniaturized sensors for DNA, pathogens, and chemicals.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service