Skip to Content
Merck

C3161

Calcium phosphate tribasic

BioReagent, suitable for plant cell culture, powder

Synonym(s):

Calcium hydroxyphosphate, HAp, Hydroxylapatite, Tribasic calcium phosphate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
[Ca5(OH)(PO4)3]x
CAS Number:
Molecular Weight:
502.31
NACRES:
NA.72
PubChem Substance ID:
UNSPSC Code:
10171502
EC Number:
235-330-6
MDL number:
Form:
powder, solid
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


product line

BioReagent

Quality Segment

form

powder, solid

technique(s)

cell culture | plant: suitable

mp

1100 °C (lit.)

application(s)

agriculture

SMILES string

[Ca++].[Ca++].[Ca++].[Ca++].O[Ca+].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O

InChI

1S/5Ca.3H3O4P.H2O/c;;;;;3*1-5(2,3)4;/h;;;;;3*(H3,1,2,3,4);1H2/q5*+2;;;;/p-10

InChI key

XYJRXVWERLGGKC-UHFFFAOYSA-D

Application

Calcium phosphate tribasic (C3161) is plant cell culture tested (0.2 mg/ml) and is appropriate for use in plant cell culture experiments. Calcium phosphate tribasic is utilized to engineer new biomaterials for applications such as bone grafts and fillers.

Preparation Note

Precipitated to reduce trace impurities.

Other Notes

Approx. Ca10(PO4)6(OH)2


Still not finding the right product?

Explore all of our products under Calcium phosphate tribasic


Storage Class

13 - Non Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library