Merck
  • The in vivo effect of esculetin ointment and esculetin-mixed Zyderm for Zyderm.
The in vivo effect of esculetin ointment and esculetin-mixed Zyderm for Zyderm.
Plastic and reconstructive surgery (2014-07-17)
Hisayo Yamaoka, Satoru Nishizawa, Masateru Matsui, Koichi Gonda, Shinichi Hirabayashi, Kazuto Hoshi, Keiko Yamaoka
ABSTRACT
Injectable collagen is often used for treatment of wrinkles or scars in cosmetic surgery. However, it is degraded within a short period after subcutaneous injection. The authors aimed to achieve a long-lasting effect of the filler with a new collagenase inhibitor, esculetin (6,7-dihydroxy-2H-chromen-2-one). Nude mice were divided into two study groups and a control group (35 mg cattle collagen): (1) those implanted with Zyderm 0.3 g subcutaneously into the dorsal region followed by daily topical application of 5% esculetin ointment (0.5 g/day) to the skin of the implanted area (the 5% esculetin ointment group), and (2) those implanted with a mixture of Zyderm 0.3 g and esculetin (1 to 4 mM) (the esculetin-mixed Zyderm groups). In each group, Zyderm was removed at different time points to measure the wet weight and hydroxyproline level. Furthermore, each removed Zyderm specimen was sectioned for histologic examination with Azan staining and immunostaining. In the esculetin ointment group and the 2 mM esculetin-mixed Zyderm group, the hydroxyproline levels at 30, 60, and 90 days were significantly higher than those in the control group, suggesting that esculetin suppresses the biodegradation of Zyderm. There was no significant difference in hydroxyproline level between the esculetin ointment group and the 2 mM esculetin-mixed Zyderm group; biodegradation occurred to a similar extent with either method of application. An atelocollagen implant is used as a safe and effective scaffold material for tissue regeneration. Future applications of the present study are expected.
Sigma-Aldrich
Collagen, Type I solution from rat tail, BioReagent, suitable for cell culture, sterile-filtered
Sigma-Aldrich
Collagen from rabbit skin, Bornstein and Traub Type I, powder
Sigma-Aldrich
Collagen from rat tail, Bornstein and Traub Type I, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from bovine nasal septum, Bornstein and Traub Type II, powder
Sigma-Aldrich
Collagen from Engelbreth-Holm-Swarm murine sarcoma basement membrane, Type IV (Miller), lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from bovine tracheal cartilage, Bornstein and Traub Type II, powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type III (Sigma Type X), powder
Sigma-Aldrich
6,7-Dihydroxycoumarin, 98%
Sigma-Aldrich
Collagen Type IV from human cell culture, Bornstein and Traub Type IV, 0.3 mg/mL, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
Collagen human, Bornstein and Traub Type I, acid soluble, powder, ~95% (SDS-PAGE)
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type I (Sigma Type VIII), powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
Collagen from chicken sternal cartilage, Type II (Miller), powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, (0.1% solution in 0.1 M acetic acid), aseptically processed, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, solid, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type V (Sigma Type IX), powder
Sigma-Aldrich
Collagen from bovine achilles tendon, powder, suitable for substrate for collagenase
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, solution, suitable for cell culture, High Performance
Esculetin, European Pharmacopoeia (EP) Reference Standard