Skip to Content
Merck
All Photos(2)

Key Documents

H4417

Sigma-Aldrich

Human Collagen Type IV

from human placenta, liquid, High Performance

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
41106305
NACRES:
NA.51

Product Name

Collagen from human placenta, Bornstein and Traub Type IV, solution, suitable for cell culture, High Performance

biological source

human placenta

Quality Level

form

solution

technique(s)

cell culture | mammalian: suitable

surface coverage

<5 μg/cm2

impurities

Endotoxin, tested
HIV, hepatitis B and hepatitis C, none detected

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

Type IV collagen has been found to play a key role in angiogenesis, neurological diseases and metastasis. Collagen from human placenta has been used to assess the bioelectric effects of quinine on airway epithelial cells, to study the selective toxicity of engineered lentvirus lytic peptides and in a particle aggregation assay for the rapid detection of fibronectin, fibrinogen and collagen receptors on Staphylococcus aureus. It has also been used in a magnesium-dependent, collagen-binding assay during characterization of human lung tumor-associated antigens.

Biochem/physiol Actions

During development, collagen IV is ubiquitously distributed in BMs. During the maturation process, this network gets partially replaced in a remarkably tissue specific manner, defining BM structure and function. Collagen IV has been shown to bind to platelets, hepatocytes, keratinocytes, endothelial, mesangial, pancreatic cells and some tumor cells.


Tissue injury in the autoimmune disease Goodpasture syndrome is due to pathogenic autoantibodies targeting the Collagen IV α3 chain . Mutations in COL4A5 are associated with Alport syndrome.

Components

All collagen molecules are composed of three polypeptide chains arranged in a triple helical conformation, with a primary structure that is mostly a repeating motif with glycine in every third position and proline or 4-hydroxyproline frequently preceding the glycine residue. Type IV collagen occurs only in the basement membranes and contains up to six genetically distinct a-chains.

Caution

Store this product at -20°C. It will retain activity for 2 years under these conditions.

Preparation Note

This product is supplied as a solution in 0.5 M acetic acid. It can be used to coat tissue-culture surfaces to promote cell attachment. The amount used in these applications is dependent on the cell type, plastic ware and incubation time. A recommended amount is 0.5-5 μg/cm2. It was prepared by pepsin extraction and chromatographic purification.

Other Notes

Collagen is classified into a number of structurally and genetically distinct types. We use the nomenclature proposed by Bornstein and Traub. Be wary of confusing Sigma-type designations with recognized collagen classification types.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

V V Vereshchaka
Fiziolohichnyi zhurnal (Kiev, Ukraine : 1994), 59(1), 97-109 (2013-05-30)
It was examined 120 persons of both sexes who had some problems with face skin. It was investigated the degree of face age changes in persons with the symptoms of skin slacses and practically healthy persons of different ages. Some
Simon Holland et al.
Current opinion in ophthalmology, 24(4), 302-309 (2013-05-18)
Topography-guided laser refractive surgery regularizes the front corneal surface irregularities to achieve the desired refractive outcome. This is particularly applicable in highly aberrated corneas, where wavefront aberrometry is often not possible. This article aims to review the recently published results
Robert Zimmermann et al.
Clinical laboratory, 59(5-6), 629-638 (2013-07-20)
The question of whether novel instruments such as multiple electrode aggregometry (MEA) can be used for measurement of the effects of nitric oxide (NO) on platelets (PLTs) has not been examined. Therefore, we compared the effects of NO concentrations (1
M V Ghica et al.
Die Pharmazie, 68(5), 340-348 (2013-06-28)
An important aspect to be considered in the healing of acute or chronic cutaneous wounds is the associated potential infection management. Collagen, the most abundant protein of the extracellular matrix, with proven properties in wounds healing and tissues regeneration, is
V K Il'in et al.
Vestnik Rossiiskoi akademii meditsinskikh nauk, (2)(2), 56-62 (2013-07-04)
Dysbiotic shifts in intestinal and pharyngeal microflora were studied in 22 normal volunteers in 9-, 14-, 105- and 520-d chamber experiments simulating some of the spaceflight factors. Two preparations were administered to prevent pharyngeal and intestinal dysbiosis: oral dry probiotic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service