Skip to Content
Merck

C5608

Collagen from rabbit skin

Bornstein and Traub Type I, powder

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12352202
EC Number:
232-697-4
NACRES:
NA.61
MDL number:
Form:
powder
Biological source:
rabbit skin
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit skin

Quality Segment

description

acid soluble

form

powder

technique(s)

cell culture | stem cell: suitable

solubility

aqueous acid: ≤10 mg/mL, aqueous sodium acetate buffer: soluble (0.01-0.5M)

UniProt accession no.

storage temp.

2-8°C

Gene Information

rabbit ... COL1A2(100008997)

General description

Collagen is classified into a number of structurally and genetically distinct types. We use the nomenclature proposed by Bornstein and Traub. Do not confuse Sigma type designations with recognized collagen classification types.

Application

Collagen-type 1 may be used in research of Idiopathic pulmonary fibrosis (IPF). Robust expression of collagen-type 1 is one distinctive feature of IPF. Additionally, collagen-type 1 has been used in studies on the effect of endoplasmic reticulum (ER) stress from IPF on myofibroblastic differentiation of lung fibroblasts. Collage-type 1 soluble in acidic solution produces three dimensional scaffolding useful in bioengineering and cell culture applications where biomaterials are needed to replace native collagen extracellular matrices.

Collagen Type I has been used as a scaffold for the growth in vitro of stem cells in a wide variety of biomaterial engineering studies.

Preparation Note

Prepared with modification of Gallop, P.M.


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library