Merck
  • The inhibitory effect of IFN-γ on protease HTRA1 expression in rheumatoid arthritis.
The inhibitory effect of IFN-γ on protease HTRA1 expression in rheumatoid arthritis.
Journal of immunology (Baltimore, Md. : 1950) (2014-06-08)
Yuzhu Hou, Haijiang Lin, Linnan Zhu, Zhaoting Liu, Fanlei Hu, Jianfeng Shi, Tao Yang, Xiaoyun Shi, Huifang Guo, Xiaotian Tan, Lianfeng Zhang, Qiang Wang, Zhanguo Li, Yong Zhao
ABSTRACT
The high temperature requirement A1 (HTRA1) is a potent protease involved in many diseases, including rheumatoid arthritis (RA). However, the regulatory mechanisms that control HTRA1 expression need to be determined. In this study, we demonstrated that IFN-γ significantly inhibited the basal and LPS-induced HTRA1 expression in fibroblasts and macrophages, which are two major cells for HTRA1 production in RA. Importantly, the inhibitory effect of IFN-γ on HTRA1 expression was evidenced in collagen-induced arthritis (CIA) mouse models and in human RA synovial cells. In parallel with the enhanced CIA incidence and pathological changes in IFN-γ-deficient mice, HTRA1 expression in the joint tissues was also increased as determined by real-time PCR and Western blots. IFN-γ deficiency increased the incidence of CIA and the pathological severity in mice. Neutralization of HTRA1 by Ab significantly reversed the enhanced CIA frequency and severity in IFN-γ-deficient mice. Mechanistically, IFN-γ negatively controls HTRA1 expression through activation of p38 MAPK/STAT1 pathway. Dual luciferase reporter assay and chromatin immunoprecipitation analysis showed that STAT1 could directly bind to HTRA1 promoter after IFN-γ stimulation. This study offers new insights into the molecular regulation of HTRA1 expression and its role in RA pathogenesis, which may have significant impact on clinical therapy for RA and possibly other HTRA1-related diseases, including osteoarthritis, age-related macular degeneration, and cancer.
Sigma-Aldrich
Anti-HtrA1 (N-term) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Collagen, Type I solution from rat tail, BioReagent, suitable for cell culture, sterile-filtered
Sigma-Aldrich
Collagen from rat tail, Bornstein and Traub Type I, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from bovine tracheal cartilage, Bornstein and Traub Type II, powder
Sigma-Aldrich
Collagen from rabbit skin, Bornstein and Traub Type I, powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type III (Sigma Type X), powder
Sigma-Aldrich
Collagen from bovine nasal septum, Bornstein and Traub Type II, powder
Sigma-Aldrich
Collagen from Engelbreth-Holm-Swarm murine sarcoma basement membrane, Type IV (Miller), lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
U0126 ethanolate, ≥98% (HPLC), powder
Sigma-Aldrich
LY 294002, LY294002, CAS 154447-36-6, is a cell-permeable, potent, reversible, and specific inhibitor of PI 3-kinase ((IC₅₀ = 1.4 µM). Acts on the ATP-binding site.
Sigma-Aldrich
Collagen Type IV from human cell culture, Bornstein and Traub Type IV, 0.3 mg/mL, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type V (Sigma Type IX), powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type I (Sigma Type VIII), powder
Sigma-Aldrich
Collagen from bovine achilles tendon, powder, suitable for substrate for collagenase
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, (0.1% solution in 0.1 M acetic acid), aseptically processed, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, solid, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen human, Bornstein and Traub Type I, acid soluble, powder, ~95% (SDS-PAGE)
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
SB 203580, SB 203580, CAS 152121-47-6, is a highly specific, potent, cell-permeable, selective, reversible, and ATP-competitive inhibitor of p38 MAP kinase (IC₅₀ = 34 nM in vitro, 600 nM in cells).
Sigma-Aldrich
SP600125, ≥98% (HPLC)
Sigma-Aldrich
Collagen from chicken sternal cartilage, Type II (Miller), powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, solution, suitable for cell culture, High Performance