Merck
  • Expression, purification, characterization, and solution nuclear magnetic resonance study of highly deuterated yeast cytochrome C peroxidase with enhanced solubility.
Expression, purification, characterization, and solution nuclear magnetic resonance study of highly deuterated yeast cytochrome C peroxidase with enhanced solubility.
Biochemistry (2013-03-23)
Alexander N Volkov, Alexandre Wohlkonig, Sameh H Soror, Nico A J van Nuland
ABSTRACT
Here we present the preparation, biophysical characterization, and nuclear magnetic resonance (NMR) spectroscopy study of yeast cytochrome c peroxidase (CcP) constructs with enhanced solubility. Using a high-yield Escherichia coli expression system, we routinely produced uniformly labeled [(2)H,(13)C,(15)N]CcP samples with high levels of deuterium incorporation (96-99%) and good yields (30-60 mg of pure protein from 1 L of bacterial culture). In addition to simplifying the purification procedure, introduction of a His tag at either protein terminus dramatically increases its solubility, allowing preparation of concentrated, stable CcP samples required for multidimensional NMR spectroscopy. Using a range of biophysical techniques and X-ray crystallography, we demonstrate that the engineered His tags neither perturb the structure of the enzyme nor alter the heme environment or its reactivity toward known ligands. The His-tagged CcP constructs remain catalytically active yet exhibit differences in the interaction with cytochrome c, the physiological binding partner, most likely because of steric occlusion of the high-affinity binding site by the C-terminal His tag. We show that protein perdeuteration greatly increases the quality of the double- and triple-resonance NMR spectra, allowing nearly complete backbone resonance assignments and subsequent study of the CcP by heteronuclear NMR spectroscopy.
Sigma-Aldrich
Cytochrome c from equine heart, BioUltra, ≥99% (SDS-PAGE), powder, suitable for mammalian cell culture
Sigma-Aldrich
Cytochrome c from bovine heart, ≥95% based on Mol. Wt. 12,327 basis
Sigma-Aldrich
Cytochrome c from equine heart, ≥95% based on Mol. Wt. 12,384 basis
Sigma-Aldrich
Cytochrome c from equine heart, ≥95% (SDS-PAGE)
Sigma-Aldrich
Cytochrome c from equine heart, BioReagent, suitable for GFC marker
Sigma-Aldrich
Cytochrome c from pigeon breast muscle, ≥95% based on Mol. Wt. 12,173 basis
Sigma-Aldrich
Cytochrome c from bovine heart, ≥95% based on Mol. Wt. 12,327 basis, powder, suitable for mammalian cell culture
Sigma-Aldrich
Cytochrome c from Saccharomyces cerevisiae, ≥85% based on Mol. Wt. 12,588 basis
Sigma-Aldrich
ProteoMass Cytochrome c MALDI-MS Standard, vial of 10 nmol, (M+H+) 12,361.96 Da by calculation