Skip to Content
Merck

C7150

Cytochrome c from equine heart

BioReagent, suitable for GFC marker

Synonym(s):

Cytochrome c from horse heart

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
23151817
EC Number:
232-700-9
NACRES:
NA.56
MDL number:
Form:
powder
Mol wt:
~12,400
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


product line

BioReagent

Quality Segment

form

powder

mol wt

~12,400

packaging

vial of ≥10 mg

suitability

suitable for GFC marker

storage temp.

−20°C

Application

Cytochrome c is primarily known as an electron-carrying mitochondrial protein and is generally regarded as a universal catalyst of respiration. Cytochrome c can be used as a gel filtration molecular weight marker in gel filtration chromatography and protein chromatography and can be used to study cytochrome c control proteins, electron transport and cellular respiration.
The specific sites and extent of oxidation in horse cytochrome c treated with H2O2 and UV were characterized. It was suggested that the state of these sites could be used as a biomarker for the oxidative environment in a cell.

Biochem/physiol Actions

Cytochrome c is primarily known as an electron-carrying mitochondrial protein. The transition of cytochrome c between the ferrous and ferric states within the cell makes it an efficient biological electron-transporter and it plays a vital role in cellular oxidations in both plants and animals. It is generally regarded as a universal catalyst of respiration, forming an essential electron-bridge between the respirable substrates and oxygen

Other Notes



Still not finding the right product?

Explore all of our products under Cytochrome c from equine heart


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library