Merck
Placental transfer of IgG subclasses in a Japanese population.
Pediatrics international : official journal of the Japan Pediatric Society (2000-09-15)
S Hashira, S Okitsu-Negishi, K Yoshino
ABSTRACT
Maternal immunoglobulin G (IgG), transferred across the placenta to the fetus during intrauterine life, is an important component of the neonatal immunological defence mechanisms against infection. There is controversy with respect to differences in placental transfer of the different IgG subclasses, and no definite data are available on a Japanese population. Therefore, we investigated placental transfer of IgG subclasses in a Japanese population. A total of 228 matched pairs of cord and maternal serum samples (20-42 weeks gestation) were assayed for each IgG subclass by an enzyme-linked immunosorbent assay. The mean values and hierarchy of cord/maternal concentration ratios of IgG subclasses at 40 weeks gestation were as follows: IgG1(1.47) > IgG3(1.17) = IgG4(1.15) > IgG2(0.80). The cord/maternal concentration ratios of all IgG subclasses were positively correlated to gestational age. The mean ratios for IgG1 and IgG4 nearly reached a plateau at 39 and 37 weeks gestation, respectively, while those for IgG2 and IgG3 increased until 41 weeks gestation. The ratios of all IgG subclasses for full-term deliveries were reciprocally correlated to the respective maternal IgG subclass serum levels. The results suggest that although all four IgG subclasses are actively transferred across the placenta, the efficiency of their transfer ranks in the order IgG1 > IgG3 = IgG4 > IgG2. The different results as to placental transfer of IgG subclasses in the literature might be due, at least in part, to different maternal IgG subclass serum levels in the populations studied.
Sigma-Aldrich
IgG from goat serum, technical grade, ≥80% (SDS-PAGE), buffered aqueous solution
Sigma-Aldrich
IgG from sheep serum, technical grade, ≥80% (SDS-PAGE), buffered aqueous solution
Sigma-Aldrich
IgG1, Kappa from murine myeloma, clone MOPC 21, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
IgG from sheep serum, reagent grade, ≥95% (SDS-PAGE), essentially salt-free, lyophilized powder
Sigma-Aldrich
IgG from porcine serum, reagent grade, ≥95% (SDS-PAGE), essentially salt-free, lyophilized powder
Sigma-Aldrich
IgG from goat serum, reagent grade, ≥95% (SDS-PAGE), essentially salt-free, lyophilized powder
Sigma-Aldrich
IgG1, Kappa from murine myeloma, clone MOPC 31C, ascites fluid, lyophilized powder
Sigma-Aldrich
IgG1−FITC Isotype Control from murine myeloma, clone MOPC 21, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
IgG1, Kappa from murine myeloma, clone MOPC 21, ascites fluid, lyophilized powder
Sigma-Aldrich
IgG3, Kappa from human myeloma plasma, purified immunoglobulin, >90% (microfluidic capillary gel electrophoresis)
Sigma-Aldrich
IgG2, Kappa from human myeloma plasma, purified immunoglobulin, >95% (SDS-PAGE)
Sigma-Aldrich
Anti-human IgG (1mg) KC, from mouse
Sigma-Aldrich
Human IgG, Fab Fragment, Plasma
Sigma-Aldrich
IgG3, Kappa from murine myeloma, clone DX, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
IgG3, Lambda from human myeloma plasma, purified immunoglobulin, >90% (microfluidic capillary gel electrophoresis)
Sigma-Aldrich
IgG1, Kappa from human myeloma plasma, purified immunoglobulin, >95% (SDS-PAGE)
Sigma-Aldrich
IgG1, Lambda from human myeloma plasma, purified immunoglobulin, >95% (SDS-PAGE)
Sigma-Aldrich
Immunoglobulin G from human serum, ≥95% (GE)
Sigma-Aldrich
IgG3, Lambda from murine myeloma, clone Y5606, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
IgG from human serum, reagent grade, ≥95% (HPLC), buffered aqueous solution
Sigma-Aldrich
IgG from rabbit serum, reagent grade, ≥95% (SDS-PAGE), essentially salt-free, lyophilized powder
Sigma-Aldrich
IgG from rabbit serum, technical grade, ≥80% (SDS-PAGE), buffered aqueous solution
Sigma-Aldrich
IgG from mouse serum, reagent grade, ≥95% (SDS-PAGE), lyophilized powder
Sigma-Aldrich
IgG from rat serum, reagent grade, ≥95% (SDS-PAGE), essentially salt-free, lyophilized powder
Sigma-Aldrich
IgG from bovine serum, reagent grade, ≥95% (SDS-PAGE), essentially salt-free, lyophilized powder
Sigma-Aldrich
IgG from human serum, technical grade, ≥80% (SDS-PAGE), buffered aqueous solution
Sigma-Aldrich
IgG from human serum, reagent grade, ≥95% (SDS-PAGE), essentially salt-free, lyophilized powder
Sigma-Aldrich
IgG2a, Lambda from murine myeloma, clone HOPC-1, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
IgG from mouse serum, technical grade, ≥80% (SDS-PAGE), buffered aqueous solution
Sigma-Aldrich
IgG from rat serum, technical grade, ≥80% (SDS-PAGE), buffered aqueous solution