Skip to Content
Merck
All Photos(4)

Documents

I5506

Sigma-Aldrich

IgG from bovine serum

reagent grade, ≥95% (SDS-PAGE), essentially salt-free, lyophilized powder

Synonym(s):

Bovine IgG

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

conjugate

unconjugated

grade

reagent grade

Assay

≥95% (SDS-PAGE)

form

essentially salt-free, lyophilized powder

storage temp.

2-8°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

IgG antibody subtype is the most abundant serum immunoglobulins of the immune system. It is secreted by B cells and is found in blood and extracellular fluids and provides protection from infections caused by bacteria, fungi and viruses. Maternal IgG is transferred to fetus through the placenta that is vital for immune defence of the neonate against infections
Bovine IgG is purified from pooled normal bovine serum by fractionation and ion-exchange chromatography.

Application

Purified bovine IgG may be used as a reference antigen, standard, blocking agent, or coating protein in a variety of immunoassays including ELISA, dot immunobinding, Western immunoblotting, immunodiffusion, and immunoelectrophoresis. Other applications include starting materials for the preparation of immunogens and solid phase immunoadsorbents. Bovine IgG was used as control in electrophoresis and in liquid chromatography.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Audrey L Shaw et al.
Clinical and experimental gastroenterology, 9, 365-375 (2016-12-17)
Previous studies have shown that oral administration of bovine immunoglobulin protein preparations is safe and provides nutritional and intestinal health benefits. The purpose of this study was to evaluate the plasma amino acid response following a single dose of serum-derived
Diane Lee et al.
F1000Research, 8, 357-357 (2019-08-29)
The epithelial lining of the lung is often the first point of interaction between the host and inhaled pathogens, allergens and medications. Epithelial cells are therefore the main focus of studies which aim to shed light on host-pathogen interactions, to
Leyton Gapper
Journal of AOAC International, 96(5), 1026-1032 (2013-11-29)
Nine laboratories participated in an AOAC collaborative study to determine bovine immunoglobulin G (IgG) levels in selected dairy powders and dietary supplements by surface plasmon resonance (SPR) methodology. Each sample matrix was dissolved in buffer and suitably diluted to fit
Patrycja Oberska et al.
International journal of molecular sciences, 25(3) (2024-02-10)
The increasing incidence of male infertility in humans and animals creates the need to search for new factors that significantly affect the course of reproductive processes. Therefore, the aim of this study was to determine the temporospatial expression of aquaglyceroporins
H G Folkesson et al.
Journal of applied physiology (Bethesda, Md. : 1985), 71(3), 1106-1111 (1991-09-01)
The passage of different-sized marker molecules over the lower respiratory tract into the blood circulation during pulmonary inflammation induced by dextran, endotoxin [i.e., lipopolysaccharide from Escherichia coli (LPS)], or ferritin was assessed in the rat. Bovine immunoglobulin G (BIgG, mol

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service