Skip to Content
Merck

Skip To

I5131

IgG from sheep serum

reagent grade, ≥95% (SDS-PAGE), essentially salt-free, lyophilized powder

Synonym(s):

Sheep IgG

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
12352203
NACRES:
NA.46
MDL number:
Conjugate:
unconjugated
Species reactivity:
-
Application:
Citations:
28
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

sheep

Quality Segment

conjugate

unconjugated

antibody form

purified immunoglobulin

grade

reagent grade

assay

≥95% (SDS-PAGE)

form

essentially salt-free, lyophilized powder

isotype

IgG

solubility

saline: 1 mg/mL at -20 °C (Repeated freezing and thawing is not recommended.)

storage temp.

2-8°C

General description

IgG consists of four isotypes IgG1, IgG2, IgG3 and IgG4. It contains four polypeptide chains, made of two identical 50 kDa γ heavy (H) chains and two identical 25 kDa κ or λ light (L) chains, which are connected by inter-chain disulfide bonds.[1]

Application

Purified IgG may be used as a reference antigen, standard, blocking agent or coating protein in a variety of immunoassays including ELISA,[2] dot immunobinding, Western immunoblotting, immunodiffusion, immunoprecipitation[3][4] and immunoelectrophoresis. Other applications include starting materials for the preparation of immunogens and solid-phase immunoadsorbents.

Biochem/physiol Actions

IgG mediates transplacental transport and participates in the secondary immune response. IgG activates the classical pathway of the complement system. It neutralizes virus particles and toxins. IgG plays a key role in antibody dependent cell-mediated cytotoxicity (ADCC) and phagocytosis.[1] It might be linked to allergy. It has the longest serum half life.
IgG antibody subtype is the most abundant of serum immunoglobulins of the immune system. It is secreted by B cells and is found in blood and extracellular fluids and provides protection from infections caused by bacteria, fungi and viruses. Maternal IgG is transferred to fetus through the placenta that is vital for immune defense of the neonate against infections.[5]

Analysis Note

Protein concentration should be determined after reconstitution by absorbance at 280 nm (E1%/280 = 14).

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
I8265I5006I5381
antibody form

purified immunoglobulin

antibody form

-

antibody form

-

antibody form

-

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

biological source

sheep

biological source

-

biological source

-

biological source

-

form

essentially salt-free, lyophilized powder

form

buffered aqueous solution

form

essentially salt-free, lyophilized powder

form

lyophilized powder

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

2-8°C

isotype

IgG

isotype

-

isotype

-

isotype

-


Still not finding the right product?

Explore all of our products under IgG from sheep serum


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters