Skip to Content
Merck
All Photos(1)

Key Documents

48670

Supelco

N-Nitrosodimethylamine (NDMA) solution

certified reference material, 200 μg/mL in methanol

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12000000
Pricing and availability is not currently available.

grade

certified reference material
TraceCERT®

Quality Level

Agency

EPA TO-7

product line

TraceCERT®

CofA

current certificate can be downloaded

feature

standard type calibration

packaging

ampule of 1 mL

concentration

200 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

single component solution

storage temp.

2-30°C

SMILES string

N(N=O)(C)C

InChI

1S/C2H6N2O/c1-4(2)3-5/h1-2H3

InChI key

UMFJAHHVKNCGLG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

N-Nitrosodimethylamine is a toxic type of N-nitrosamine[1], which can be commonly synthesized in vivo by intestinal microbiota type of microorganisms.[2] It can undergo reduction to form dimethylamine using zero-valent metal ions like Fe and Zn in the presence of Cu(II) ions.[1]

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Carc. 1B - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jin Woong Kim et al.
Journal of magnetic resonance imaging : JMRI, 53(2), 554-563 (2020-07-03)
Liver fibrosis is characterized by the excessive accumulation of extracellular matrix proteins. Electrical conductivity imaging at low frequency can provide novel contrast because the contrast mechanisms originate from the changes in the concentration and mobility of ions in the extracellular
Lactic acid bacteria directly degrade N-nitrosodimethylamine and increase the nitrite-scavenging ability in kimchi
Kim H-S, et al.
Food Control, 71, 101-109 (2017)
The role of Cu (II) in the reduction of N-nitrosodimethylamine with iron and zinc
Han Y, et al.
Chemosphere, 167, 171-177 (2017)
András P Keszei et al.
The American journal of clinical nutrition, 97(1), 135-146 (2012-11-30)
Dietary N-nitroso compounds and endogenous nitrosation are important carcinogenic factors, but human evidence of their role is scarce for esophageal cancer and inconsistent for gastric cancer. We studied the relation between risks of esophageal and gastric cancer subtypes and dietary
Ina Kristiana et al.
Water research, 47(2), 535-546 (2012-11-21)
N-Nitrosamines are a class of disinfection by-products (DBPs) that have been reported to be more toxic than the most commonly detected and regulated DBPs. Only a few studies investigating the formation of N-nitrosamines from disinfection of natural waters have been

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service