Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C2H6N2O
CAS Number:
Molecular Weight:
74.08
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
grade
pharmaceutical primary standard
agency
USP
form
liquid
packaging
pkg of 1 mg
manufacturer/tradename
USP
application(s)
USP Biologics
pharmaceutical (small molecules)
format
single component solution (solution in methanol)
storage temp.
−20°C
SMILES string
N(N=O)(C)C
InChI
1S/C2H6N2O/c1-4(2)3-5/h1-2H3
InChI key
UMFJAHHVKNCGLG-UHFFFAOYSA-N
Application
N-Nitrosodimethylamine (NDMA) USP reference standard is intended for use only as specificallyprescribed in the United States Pharmacopoeia.
It is also used to prepare standard, standard stock, nitrosamine RS stock, nitrosamine standards stock solution mixture, and sensitivity stock solutions to determine NDMA impurity in drug substances and drug products(valsartan, irbesartan, and losartan potassium etc.) by chromatography method according to the general chapter 〈1469〉 of United States Pharmacopeia.
It is also used to prepare standard, standard stock, nitrosamine RS stock, nitrosamine standards stock solution mixture, and sensitivity stock solutions to determine NDMA impurity in drug substances and drug products(valsartan, irbesartan, and losartan potassium etc.) by chromatography method according to the general chapter 〈1469〉 of United States Pharmacopeia.
Analysis Note
These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.
Other Notes
Sales restrictions may apply.
This product is part of USP Biologics Program.
This product is provided as delivered and specified by the USP Pharmacopoeia. For further information and support, including certificate/ product information sheets, please go to the website of the issuing Pharmacopoeia.
Still not finding the right product?
Explore all of our products under N-Nitrosodimethylamine (NDMA)
signalword
Danger
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Carc. 1B - Flam. Liq. 2 - STOT SE 1
target_organs
Eyes,Central nervous system
Storage Class
3 - Flammable liquids
wgk
WGK 3
flash_point_f
49.5 °F
flash_point_c
9.7 °C
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.


