Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C2D6N2O
CAS Number:
Molecular Weight:
80.12
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
grade
pharmaceutical primary standard
agency
USP
packaging
pkg of 1 mL
manufacturer/tradename
USP
application(s)
pharmaceutical (small molecule)
format
neat
storage temp.
−20°C
SMILES string
N(N=O)(C([2H])([2H])[2H])C([2H])([2H])[2H]
InChI
1S/C2H6N2O/c1-4(2)3-5/h1-2H3/i1D3,2D3
InChI key
UMFJAHHVKNCGLG-WFGJKAKNSA-N
General description
Nitrosamines are chemical compounds that can form during drug manufacturing and may pose health risks, including carcinogenicity.
This product is provided as delivered and specified by the issuing Pharmacopoeia. For further information and support, including certificate/ product information sheets, please go to the website of the issuing Pharmacopoeia.
This product is provided as delivered and specified by the issuing Pharmacopoeia. For further information and support, including certificate/ product information sheets, please go to the website of the issuing Pharmacopoeia.
Application
Deutero N-Nitrosodimethylamine (NDMA-d6) USP reference standard is intended for use only as specifically prescribed in the United States Pharmacopoeia.
Also used to prepare internal standard, and internal standard stock solutions to determine nitrosamine impurity in drug substances and drug products by chromatography according to general chapter <1469> of United States Pharmacopeia.
Also used to prepare internal standard, and internal standard stock solutions to determine nitrosamine impurity in drug substances and drug products by chromatography according to general chapter <1469> of United States Pharmacopeia.
Analysis Note
These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.
Other Notes
Sales restrictions may apply.
signalword
Danger
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Carc. 1B - Flam. Liq. 2 - STOT SE 1
target_organs
Eyes,Central nervous system
Storage Class
3 - Flammable liquids
wgk
WGK 3
flash_point_f
49.5 °F
flash_point_c
9.7 °C
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.


