Skip to Content
Merck
All Photos(1)

Key Documents

502138

Supelco

EPA 8270/Appendix IX Nitrosamines Mix

certified reference material, 2000 μg/mL each component in methanol

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000

grade

certified reference material
TraceCERT®

Agency

EPA 8270

product line

TraceCERT®

CofA

current certificate can be downloaded

feature

standard type calibration

packaging

ampule of 1 mL

concentration

2000 μg/mL each component in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

multi-component solution

storage temp.

-10 to -25°C

Application

EPA 8270/Appendix IX nitrosamines mix has been used as a standard solution in the determination of N-nitrosamine in water samples using gas chromatography coupled with mass spectrometry (GC-MS). It has also been used as a standard in the determination of nicotine and N-nitrosamines in indoor house dust using gas chromatography (GC).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    N-Nitrosodibutylamine
    N-Nitrosodiethylamine
    N-Nitrosodimethylamine
    N-Nitrosodiphenylamine
    N-Nitrosodi-n-propylamine
    N-Nitrosomethylethylamine
    Nitrosomorpholine
    1-Nitrosopiperidine
    1-Nitrosopyrrolidine
    See All (9)

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Carc. 1B - Flam. Liq. 2 - Skin Sens. 1 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joy Roasa et al.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment, 36(3), 396-404 (2019-02-08)
A method to detect volatile nitrosamines in meat samples was developed using headspace sampling by solid-phase microextraction (HS-SPME), with analysis by GC-MS. A 50/30 µm divinylbenzene/carboxen/polydimethylsiloxane fused silica fibre was selected to extract a total of nine volatile nitrosamines: N-nitrosodimethylamine, N-nitrosomethylethylamine
Wilson Beita-Sandí et al.
Chemosphere, 211, 1091-1097 (2018-09-19)
Cation exchange resins have proved to be efficient in removing precursors of N-nitrosodimethylamine (NDMA). NDMA is a probable human carcinogen with a calculated lifetime cancer risk of 10-6 at 0.7 ng/L in drinking water. This paper investigated the effect of pH and
Fully automated determination of N?nitrosamines in environmental waters by headspace solid?phase microextraction followed by GC?MS?MS
Llop A, et al.
Journal of Separation Science, 33, 3692-3700 (2010)
Determination of nicotine and N-nitrosamines in house dust by pressurized liquid extraction and comprehensive gas chromatography?Nitrogen chemiluminiscence detection
Ramirez N, et al.
Journal of Chromatography A, 1219, 180-187 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service