Skip to Content
Merck
All Photos(1)

Key Documents

47865-U

Supelco

Nicotinamide (Niacinamide)

analytical standard

Synonym(s):

Nicotinamide, NAM, NIC, Niacinamide, Nicotinic acid amide, Pyridine-3-carboxylic acid amide, Vitamin B3, Vitamin PP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6N2O
CAS Number:
Molecular Weight:
122.12
Beilstein:
383619
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

CofA

current certificate can be downloaded

feature

standard type water soluble vitamin

packaging

ampule of 1000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

128-131 °C (lit.)

application(s)

clinical testing

format

neat

storage temp.

2-30°C

SMILES string

NC(=O)c1cccnc1

InChI

1S/C6H6N2O/c7-6(9)5-2-1-3-8-4-5/h1-4H,(H2,7,9)

InChI key

DFPAKSUCGFBDDF-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Nicotinamide (Niacinamide), belonging to the class of water-soluble B vitamins, is typically produced from nicotinic acid (vitamin B3). It is found as a component of food products. It has been suggested for the treatment of pellagra, psoriasis, schizophrenia, and type I diabetes. It also exhibits anti-inflammatory activity.

Application

Nicotinamide (Niacinamide) may be used as a reference standard for the determination of the analyte in pharmaceutical formulations and biological fluids by chromatography, UV-visible spectrophotometry and multivariate analysis.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Nicotinamide is an amide derivative of vitamin B3 and a PARP inhibitor

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

302.0 °F - closed cup

Flash Point(C)

150 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Industrial application of green chromatography?I. Separation and analysis of niacinamide in skincare creams using pure water as the mobile phase
Yang Y, et al.
Talanta, 84(1), 169-174 (2011)
Development of a validated HPLC method for the determination of B-complex vitamins in pharmaceuticals and biological fluids after solid phase extraction
Chatzimichalakis PF, et al.
Journal of Separation Science, 27(14), 1181-1188 (2004)
Efrat Resnick et al.
Journal of the American Chemical Society, 141(22), 8951-8968 (2019-05-08)
Covalent probes can display unmatched potency, selectivity, and duration of action; however, their discovery is challenging. In principle, fragments that can irreversibly bind their target can overcome the low affinity that limits reversible fragment screening, but such electrophilic fragments were
Nicotinamide
European Pharmacopoeia Commission and European Directorate for the Quality of Medicines & Healthcare
European pharmacopoeia, 14(4), 3145-3146 (2017)
Evaluation of analytical tools and multivariate methods for quantification of co-former crystals in ibuprofen-nicotinamide co-crystals
Soares FLF and Carneiro RL
Journal of Pharmaceutical and Biomedical Analysis, 89, 166-175 (2014)

Articles

Learn to separate and quantify B vitamins and preservatives in capsules with HPLC-UV and Purospher® STAR RP-8 column. Validated for precision.

Learn to separate and quantify B vitamins and preservatives in capsules with HPLC-UV and Purospher® STAR RP-8 column. Validated for precision.

Learn to separate and quantify B vitamins and preservatives in capsules with HPLC-UV and Purospher® STAR RP-8 column. Validated for precision.

Learn to separate and quantify B vitamins and preservatives in capsules with HPLC-UV and Purospher® STAR RP-8 column. Validated for precision.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service