Skip to Content
Merck
All Photos(1)

Key Documents

X2628

Sigma-Aldrich

Xestospongin C

film

Synonym(s):

XeC

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C28 H50 N2 O2
CAS Number:
Molecular Weight:
446.71
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

form

film

color

colorless

solubility

DMSO: soluble

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@]12CCCCCC[C@@]3([H])CCCN4CC[C@@]([H])(CCCCCC[C@]5([H])CCCN(CC1)[C@@]5([H])O2)O[C@@]34[H]

InChI

1S/C28H50N2O2/c1-3-7-15-25-17-21-30-20-10-14-24(28(30)31-25)12-6-2-4-8-16-26-18-22-29-19-9-13-23(11-5-1)27(29)32-26/h23-28H,1-22H2/t23-,24+,25-,26-,27+,28+/m1/s1

InChI key

PQYOPBRFUUEHRC-HCKQMYSWSA-N

General description

Synthetic form of the macrocyclic bis-1-oxaquinolizidine isolated from the Okanowan marine sponge.

Biochem/physiol Actions

Xestospongin C is a potent, reversible and membrane-permeable blocker of IP3-mediated Ca2+ release (IC50=358 nM).
Xestospongin C is a selective, reversible and membrane-permeable inhibitor of IP3 receptor. Reversibly blocks bradykinin- and carbamylcholine- Ca2+ efflux from the endoplasmic reticulum stores.

Features and Benefits

This compound is featured on the InsP3/Ryanodine Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Packaging

Package under nitrogen, tightly sealed.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Martin Prince Alphonse et al.
Journal of immunology (Baltimore, Md. : 1950), 197(9), 3481-3489 (2016-10-04)
Kawasaki disease (KD) is a multisystem vasculitis that predominantly targets the coronary arteries in children. Phenotypic similarities between KD and recurrent fever syndromes point to the potential role of inflammasome activation in KD. Mutations in NLRP3 are associated with recurrent
Hirofumi Morihara et al.
PloS one, 12(12), e0189948-e0189948 (2017-12-22)
Excessive levels of reactive oxygen species (ROS) and impaired Ca2+ homeostasis play central roles in the development of multiple cardiac pathologies, including cell death during ischemia-reperfusion (I/R) injury. In several organs, treatment with 2-aminoethoxydiphenyl borate (2-APB) was shown to have
S Miyamoto et al.
British journal of pharmacology, 130(3), 650-654 (2000-05-24)
We evaluated the role of the inositol 1,4,5-triphosphate (IP(3)) receptor-mediated Ca(2+) release on the positive inotropic effects of alpha-adrenergic stimulation using a novel, potent, selective membrane-permeable blocker of IP(3) receptor, xestospongin C. Guinea-pig papillary muscle permeabilized with saponin exhibited spontaneous
J Gafni et al.
Neuron, 19(3), 723-733 (1997-10-23)
Xestospongins (Xe's) A, C, D, araguspongine B, and demethylxestospongin B, a group of macrocyclic bis-1-oxaquinolizidines isolated from the Australian sponge, Xestospongia species, are shown to be potent blockers of IP3-mediated Ca2+ release from endoplasmic reticulum vesicles of rabbit cerebellum. XeC
Wenbin Zhong et al.
Nature communications, 7, 12702-12702 (2016-09-02)
Metabolic pathways are reprogrammed in cancer to support cell survival. Here, we report that T-cell acute lymphoblastic leukemia (T-ALL) cells are characterized by increased oxidative phosphorylation and robust ATP production. We demonstrate that ORP4L is expressed in T-ALL but not

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service