Skip to Content
Merck

D9175

Dantrolene sodium salt

Ca2+ release from sarcoplasmic reticulum inhibitor, solid

Synonym(s):

1-[(5-­(p-Nitro­phenyl)­fur­furyl­idene)­amino]­hydan­toin sodium salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C14H9N4NaO5
CAS Number:
Molecular Weight:
336.23
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
MDL number:
Form:
solid
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Dantrolene sodium salt,

form

solid

Quality Segment

originator

JHP Pharmaceuticals

SMILES string

[Na+].[H]\C(=N/N1CC(=O)N=C1[O-])c2ccc(o2)-c3ccc(cc3)[N+]([O-])=O

InChI

1S/C14H10N4O5.Na/c19-13-8-17(14(20)16-13)15-7-11-5-6-12(23-11)9-1-3-10(4-2-9)18(21)22;/h1-7H,8H2,(H,16,19,20);/q;+1/p-1/b15-7+;

InChI key

KSRLIXGNPXAZHD-HAZZGOGXSA-M

Gene Information

Biochem/physiol Actions

Inhibitor of Ca2+ release from sarcoplasmic reticulum; muscle relaxant.

Features and Benefits

This compound is featured on the InsP3/Ryanodine Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by JHP Pharmaceuticals. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.


Still not finding the right product?

Explore all of our products under Dantrolene sodium salt


Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library