Skip to Content
Merck

Skip To

SML1845

Thapsigargin

From (Thapsia garganica L seeds), ≥98% (HPLC), calcium ion pumps inhibitor, DMSO solution

Synonym(s):

CHEBI:9516, CHEMBL96926, MFCD00083511, UNII-Z96BQ26RZD, Z96BQ26RZD, [(3S,3aR,4S,6S,6aR,7S,8S,9bS)-6-acetyloxy-4-butanoyloxy-3,3a-dihydroxy-3,6,9-trimethyl-8-[(Z)-2-methylbut-2-enoyl]oxy-2-oxo-4,5,6a,7,8,9b-hexahydroazuleno[4,5-b]furan-7-yl] octanoate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C34H50O12
CAS Number:
Molecular Weight:
650.75
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
DMSO solution
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Thapsigargin Ready Made Solution, ≥98% (HPLC), 10 mM in DMSO

biological source

(Thapsia garganica L seeds)

Quality Segment

assay

≥98% (HPLC)

form

DMSO solution

concentration

10 mM in DMSO

shipped in

dry ice

storage temp.

−20°C

SMILES string

O1[C@@H]2[C@]([C@H](C[C@@]([C@H]3[C@@H]([C@H](C(=C32)C)OC(=O)\C(=C/C)\C)OC(=O)CCCCCCC)(OC(=O)C)C)OC(=O)CCC)([C@@](C1=O)(O)C)O

InChI

1S/C34H50O12/c1-9-12-13-14-15-17-24(37)43-28-26-25(20(5)27(28)44-30(38)19(4)11-3)29-34(41,33(8,40)31(39)45-29)22(42-23(36)16-10-2)18-32(26,7)46-21(6)35/h11,22,26-29,40-41H,9-10,12-18H2,1-8H3/b19-11-/t22-,26+,27-,28-,29-,32-,33+,34+/m0/s1

InChI key

IXFPJGBNCFXKPI-FSIHEZPISA-N

Application

Thapsigargin Ready Made Solution has been used as a negative control in in vivo growth and in vitro translation measurements.

Biochem/physiol Actions

Thapsigargin is an effective inhibitor of calcium ion pumps located on sarcoplasmic reticulum (SR) and endoplasmic reticulum (ER) microsomes of skeletal, cardiac, muscle and brain tissues.
Thapsigargin is an effective inhibitor of calcium ion pumps located on sarcoplasmic reticulum (SR) and endoplasmic reticulum (ER) microsomes of skeletal, cardiac, muscle and brain tissues. The inhibition of calcium ion pumps causes intracellular increase of calcium ion levels. It was found that thapsigargin at 100 nM concentration effectively inhibited the SR Ca2+- adenosine triphosphatase (ATPase) in cardiac and skeletal muscles. Thapsigargin was also reported to be effective in blocking autophagy by interfering with the autophagosome-lysosome fusion.

Other Notes

Thapsigargin Ready Made Solution is provided at 10 mM concentration in DMSO. The solution can be further diluted with DMSO to a concentration of 2 mM. It is recommended to store Thapsigargin Ready Made Solution in small aliquots at -20 °C.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
586006T9033586005
description

≥98% (HPLC), 10 mM in DMSO

description

InSolution, ≥97%, tumor-promoting sesquiterpene lactone

description

≥98% (HPLC), solid film

description

Thapsigargin, CAS 67526-95-8, is a cell-permeable, tumor-promoting sesquiterpene lactone that releases calcium by non-competitvley inhibiting endoplasmic reticular Ca2+-ATPase (IC50 = 4-13 nM).

assay

≥98% (HPLC)

assay

≥97% (TLC)

assay

≥98% (HPLC)

assay

≥97% (TLC)

form

DMSO solution

form

liquid

form

solid film

form

solid

Quality Level

200

Quality Level

100

Quality Level

300

Quality Level

200

storage temp.

−20°C

storage temp.

−70°C

storage temp.

−20°C

storage temp.

−20°C

shipped in

dry ice

shipped in

dry ice

shipped in

-

shipped in

ambient

biological source

(Thapsia garganica L seeds)

biological source

-

biological source

plant (Thapsia garganica)

biological source

-


Still not finding the right product?

Explore all of our products under Thapsigargin Ready Made Solution


Storage Class

10 - Combustible liquids

WGK

WGK 2

Flash Point (°F)

188.6 °F - closed cup

Flash Point (°C)

87 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters