Skip to Content
Merck
All Photos(1)

Key Documents

SML1531

Sigma-Aldrich

Treprostinil

≥98% (HPLC)

Synonym(s):

2-[[(1R,2R,3aS,9aS)-2,3,4,9,9a-hexahydro-2-hydroxy-1-[(3S)-3-hydroxyoctyl]-1H-benz[f]inden-5-yl]oxy]-acetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H34O5
CAS Number:
Molecular Weight:
390.51
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D 40 to 50°, c = 0.5 in methanol

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

[H][C@@]12[C@@](C[C@@H](O)[C@@H]2CC[C@@H](O)CCCCC)([H])CC3=C(OCC(O)=O)C=CC=C3C1

InChI

1S/C23H34O5.Na/c1-2-3-4-7-17(24)9-10-18-19-11-15-6-5-8-22(28-14-23(26)27)20(15)12-16(19)13-21(18)25;/h5-6,8,16-19,21,24-25H,2-4,7,9-14H2,1H3,(H,26,27);/q;+1/p-1/t16-,17-,18+,19-,21+;/m1./s1

InChI key

IQKAWAUTOKVMLE-DQHZMXPSSA-M

Biochem/physiol Actions

Treprostinil elicits a longer half-life when compared with iloprost. It mediates inhibition of transforming growth factor-β (TGF-β) and nuclear factor-κB (NF-κB) signaling pathways. Treprostinil effectively reduces right ventricular systolic pressure and inhibits the recruitment of circulating fibrocyte.
Treprostinil is a stable synthetic analog of prostacyclin (prostaglandin PGI2), an IP receptor agonist used clinically for the treatment of pulmonary arterial hypertension. It is a potent vasodilator with antithrombotic, antiproliferative and anti-inflammatory properties.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Robert Roscigno et al.
Pulmonary circulation, 10(4), 2045894020971509-2045894020971509 (2020-12-08)
A dry-powder inhaled formulation of treprostinil (LIQ861) produced using PRINT® technology offers a substantial advantage over current nebulized therapy. Treprostinil is a synthetic prostacyclin analogue that is currently approved for inhalation administration to patients with pulmonary arterial hypertension via nebulized
Ming-Kai Tsai et al.
Journal of investigative medicine : the official publication of the American Federation for Clinical Research, 62(2), 332-339 (2014-01-10)
Inflammation plays critical roles in atherosclerosis. Chemokines are responsible for leukocyte trafficking and involve in inflammatory diseases. Macrophage inflammatory protein 1α (MIP-1α) has been implicated in atherosclerotic lesion formation. Prostaglandin I2 (PGI2) analog, used in pulmonary hypertension, has been reported
Alessio Coi et al.
Orphanet journal of rare diseases, 16(1), 90-90 (2021-02-19)
Systemic Sclerosis (SSc) is a chronic autoimmune disease with a complex pathogenesis that includes vascular injury, abnormal immune activation, and tissue fibrosis. We provided a complete epidemiological characterization of SSc in the Tuscany region (Italy), considering prevalence and incidence, survival
Aaron Waxman et al.
The New England journal of medicine, 384(4), 325-334 (2021-01-14)
No therapies are currently approved for the treatment of pulmonary hypertension in patients with interstitial lung disease. The safety and efficacy of inhaled treprostinil for patients with this condition are unclear. We enrolled patients with interstitial lung disease and pulmonary
R James White et al.
American journal of respiratory and critical care medicine, 201(6), 707-717 (2019-11-26)
Rationale: Oral treprostinil improves exercise capacity in patients with pulmonary arterial hypertension (PAH), but the effect on clinical outcomes was unknown.Objectives: To evaluate the effect of oral treprostinil compared with placebo on time to first adjudicated clinical worsening event in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service