Skip to Content
Merck
All Photos(1)

Key Documents

30165

Sigma-Aldrich

Tirofiban

≥98.5% (HPLC)

Synonym(s):

(2S)-2-(Butylsulfonylamino)-3-{4-[4-(4-piperidyl)butoxy}phenyl propanoic acid, N-(Butylsulfonyl)-O-[4-(4-piperidinyl)butyl]-L-tyrosine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H36N2O5S
CAS Number:
Molecular Weight:
440.60
Beilstein:
6182267
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥98.5% (HPLC)

form

powder

SMILES string

CCCCS(=O)(=O)N[C@@H](Cc1ccc(OCCCCC2CCNCC2)cc1)C(O)=O

InChI

1S/C22H36N2O5S/c1-2-3-16-30(27,28)24-21(22(25)26)17-19-7-9-20(10-8-19)29-15-5-4-6-18-11-13-23-14-12-18/h7-10,18,21,23-24H,2-6,11-17H2,1H3,(H,25,26)/t21-/m0/s1

InChI key

COKMIXFXJJXBQG-NRFANRHFSA-N

Gene Information

Application


  • The efficacy and safety of continuous intravenous tirofiban for acute ischemic stroke patients treated by endovascular therapy: a meta-analysis.: This meta-analysis assesses the safety and efficacy of continuous intravenous tirofiban in patients undergoing endovascular therapy for acute ischemic stroke, showing promising results for improved outcomes (Wang et al., 2024).

  • Initial Experience with a New Self-Expanding Open-Cell Stent System with Antithrombotic Hydrophilic Polymer Coating (pEGASUS Stent) in the Treatment of Wide-Necked Intracranial Aneurysms.: This study reports on the initial use of a new stent system coated with tirofiban in treating intracranial aneurysms, demonstrating enhanced safety and efficacy (Boxberg et al., 2024).

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marco Valgimigli et al.
JACC. Cardiovascular interventions, 5(3), 268-277 (2012-03-24)
The authors sought to compare the effect on inhibition of platelet aggregation (IPA) of prasugrel therapy versus tirofiban bolus with or without a post-bolus short drug infusion in ST-segment elevation myocardial infarction (STEMI) patients. The degree and rapidity of IPA
Gang Luo et al.
World neurosurgery, 109, e318-e328 (2017-10-11)
Early recanalization of acute posterior circulation stroke caused by large intracranial vessel occlusion by mechanical thrombectomy with stent retrievers may improve the outcome of patients. However, evidence of patient selection is still lacking. This study investigated the prognostic factors of
Shukri W David et al.
Catheterization and cardiovascular interventions : official journal of the Society for Cardiac Angiography & Interventions, 93(5), 882-890 (2018-09-29)
In the randomized AMIHOT-II trial, supersaturated oxygen [SSO2 ] delivered into the left anterior descending (LAD) artery via an indwelling intracoronary infusion catheter following primary percutaneous coronary intervention (PCI) significantly reduced infarct size in patients with anterior ST-segment elevation myocardial
Christoph Kabbasch et al.
Journal of neurointerventional surgery, 11(8), 812-816 (2019-01-25)
Intrasaccular flow disruption represents a new paradigm in endovascular treatment of wide-necked bifurcation aneurysms. To perform a matched case-control study comparing complications and angiographic outcome using the Woven Endobridge (WEB) device and stent-assisted coiling (SAC). Sixty-six patients treated with the
Nohra Chalouhi et al.
Neurosurgery, 71(3), 710-714 (2012-06-07)
Thromboembolic complications are a major concern in stent-assisted coiling of intracranial aneurysms that may be prevented with adequate antiplatelet therapy. To assess the safety and efficacy of tirofiban in stent-assisted coiling. Two protocols were used. In the initial protocol, tirofiban

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service