Skip to Content
Merck
All Photos(1)

Key Documents

SML2653

Sigma-Aldrich

Sorafenib

≥98% (HPLC), powder, kinase inhibitor

Synonym(s):

4-[4-[[[[4-Chloro-3-(trifluoromethyl)phenyl]amino]carbonyl]amino]phenoxy]-N-methyl-2-pyridinecarboxamide, BAY 43-9006, BAY43-9006, N-[4-Chloro-3-(trifluoromethyl)phenyl]-N′-[4-[2-(N-methylcarbamoyl)-4-pyridyloxy]phenyl]urea

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H16ClF3N4O3
CAS Number:
Molecular Weight:
464.82
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Product Name

Sorafenib, ≥98% (HPLC)

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

FC(F)(F)c1c(ccc(c1)NC(=O)Nc2ccc(cc2)Oc3cc(ncc3)C(=O)NC)Cl

InChI

1S/C21H16ClF3N4O3/c1-26-19(30)18-11-15(8-9-27-18)32-14-5-2-12(3-6-14)28-20(31)29-13-4-7-17(22)16(10-13)21(23,24)25/h2-11H,1H3,(H,26,30)(H2,28,29,31)

InChI key

MLDQJTXFUGDVEO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Sorafenib (BAY 43-9006) is an orally active kinase inhibitor that exerts broad-spectrum anticancer efficacy in vitro and in vivo via targeting b-Raf, c-Raf (Raf-1), as well as several receptor tyrosine kinases involved in neovascularization and tumor progression, including vascular endothelial growth factor receptors 2/3 (VEGFR-2/Flk-1/KDR, VEGFR-3), platelet-derived growth factor receptor-beta (PDGFR-ß), Flt-3, c-KIT, FGFR-1 (Flt-2) and RET.

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Lact. - Repr. 1B - STOT RE 1

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Paul T C Wan et al.
Cell, 116(6), 855-867 (2004-03-24)
Over 30 mutations of the B-RAF gene associated with human cancers have been identified, the majority of which are located within the kinase domain. Here we show that of 22 B-RAF mutants analyzed, 18 have elevated kinase activity and signal
Jeong-Ju Yoo et al.
International journal of molecular sciences, 20(6) (2019-03-17)
This study aimed to examine whether inhibition of hexokinase (HK)-II activity enhances the efficacy of sorafenib in in-vivo models of hepatocellular carcinoma (HCC), and to evaluate the prognostic implication of HK-II expression in patients with HCC. We used 3-bromopyruvate (3-BP)
Hyewon Jang et al.
Antioxidants (Basel, Switzerland), 11(10) (2022-10-28)
Alpinumisoflavone is a natural prenylated isoflavonoid extracted from the raw fruit of Cudrania tricuspidata. Several studies have reported the beneficial characteristics of alpinumisoflavone, such as its antioxidant, anti-inflammation, anti-bacterial, osteoprotective, and neuroprotective effects. Alpinumisoflavone also has anti-cancer effects on thyroid
Xudong Gao et al.
OncoTargets and therapy, 12, 1629-1640 (2019-03-19)
Hepatocellular carcinoma (HCC) is one of the heaviest malignant burdens in China. Molecular targeting agent, sorafenib, is the main therapeutic option for antitumor therapy of advanced HCC, but it is currently too expensive for the public and its therapeutic effect
Hung Huynh et al.
International journal of oncology, 54(3), 1123-1133 (2019-02-13)
Mutations affecting the Wnt/β‑catenin pathway have been identified in 26‑40% of hepatocellular carcinoma (HCC) cases. Aberrant activation of this pathway leads to uncontrolled cell proliferation and survival. Thus, identifying Wnt/β‑catenin pathway inhibitors may benefit a subset of patients with HCC.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service