Skip to Content
Merck

Skip To

A9525

Angiotensin II human

≥93% (HPLC), powder

Synonym(s):

1-8-AngiotensinI, 5-Isoleucine-angiotensinII, Ang II, Angiotensin 2, Angiotensin II, DRVYIHPF, Hypertensin, Ile(5)-angiotensinII

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C50H71N13O12
CAS Number:
Molecular Weight:
1046.18
MDL number:
UNSPSC Code:
12352202
eCl@ss:
42020611
PubChem Substance ID:
NACRES:
NA.32
Form:
powder
Assay:
≥93% (HPLC)
Biological source:
synthetic
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

synthetic

Quality Segment

assay

≥93% (HPLC)

form

powder

storage condition

(Keep container tightly closed in a dry and well-ventilated place)

concentration

≤100%

technique(s)

cell culture | mammalian: suitable

impurities

1.2-10.4% acetate

solubility

water: 25 mg/mL, clear, colorless

UniProt accession no.

storage temp.

−20°C

SMILES string

[nH]1cncc1C[C@H](NC(=O)C(NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](N)CC(=O)O)CCCNC(=N)N)C(C)C)Cc4ccc(cc4)O)[C@H](CC)C)C(=O)N2[C@@H](CCC2)C(=O)N[C@@H](Cc3ccccc3)C(=O)O

InChI

1S/C50H71N13O12/c1-5-28(4)41(47(72)59-36(23-31-25-54-26-56-31)48(73)63-20-10-14-38(63)45(70)60-37(49(74)75)22-29-11-7-6-8-12-29)62-44(69)35(21-30-15-17-32(64)18-16-30)58-46(71)40(27(2)3)61-43(68)34(13-9-19-55-50(52)53)57-42(67)33(51)24-39(65)66/h6-8,11-12,15-18,25-28,33-38,40-41,64H,5,9-10,13-14,19-24,51H2,1-4H3,(H,54,56)(H,57,67)(H,58,71)(H,59,72)(H,60,70)(H,61,68)(H,62,69)(H,65,66)(H,74,75)(H4,52,53,55)/t28-,33-,34-,35-,36-,37-,38-,40-,41?/m0/s1

InChI key

CZGUSIXMZVURDU-GYGDIBSWSA-N

Gene Information

human ... AGT(183)

General description

Asp-Arg-Val-Tyr-Ile-His-Pro-Phe-OH · 2CH3COOH · H2O
Research area: Cell Signaling

Application

  • Angiotensin II (angII) human has been used:
  • to study its effect on interleukin-1β–mediated islet inflammation and β-cell dysfunction.
  • to determine if angII activates specific calcineurin-dependent NFAT (nuclear factor of activated T-cells) isoforms in cardiomyocytes.
  • to study the actions of systemic angII infusions on glomerular permeability in vivo.
  • to induce abdominal aortic aneurysms (AAAs) in ApoE knockout mice for experimental studies.
  • to study the effect of Ang II infusion on atrial branched-chain amino acids (BCAAs) catabolism in male C57/BL6 mice.
  • to check the effects of angII on plasma triglyceride production and its association with insulin resistance in normal rats.

Biochem/physiol Actions

Angiotensin II (angII) is an octapeptide cardiovascular hormone which affects the activities of the heart, kidney, vasculature and brain. AngII in the blood is produced from angiotensinogen via renin and angiotensin converting enzyme (ACE). It works via binding to specific receptors present on cell membranes. It is associated with vasoconstriction, induction of vascular smooth muscle cell growth, stimulation of protooncogene expression, and modulation of myocardial hypertrophy, renal failure and fibrosisAngII plays a key role in the regulation of blood pressure via the renin-angiotensin-aldosterone (RAAS) pathway.

Features and Benefits

The purity of our angiotensin II human, Ang II, is verified to assure your experimental success (HPLC and peptide content). The peptide sequence is DRVYIHPF.

Analysis Note

Peptide content not less than 75%

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
A965010385SAB4200885
description

≥93% (HPLC), powder

description

≥90% (HPLC), powder, suitable for mammalian cell culture

description

≥98% (HPCE)

description

affinity isolated antibody, buffered aqueous solution

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

-

technique(s)

-

biological source

synthetic

biological source

human

biological source

-

biological source

-

assay

≥93% (HPLC)

assay

≥90% (HPLC)

assay

≥98% (HPCE)

assay

-

concentration

≤100%

concentration

-

concentration

-

concentration

~1 mg/mL

form

powder

form

powder

form

-

form

buffered aqueous solution, liquid

Gene Information

human ... AGT(183)

Gene Information

human ... AGT(183)

Gene Information

-

Gene Information

-


Still not finding the right product?

Explore all of our products under Angiotensin II human


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

  1. What is the molar mass of human Angiotensin II (A9525)?

    1 answer
    1. The molecular weight of this material is 1046.18 g/mol.

      Helpful?

Reviews

Active Filters

  1. 1 Ratings-only review