Skip to Content
Merck
All Photos(1)

Key Documents

SML1495

Sigma-Aldrich

Suprofen

≥98% (HPLC)

Synonym(s):

(±)-Suprofen, 2-[4-(Thiophene-2-carbonyl)phenyl]propanoic acid, NSC 303611, p-(2-Thenoyl)hydratropic acid, α-Methyl-p-(2-thenoyl)phenylacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H12O3S
CAS Number:
Molecular Weight:
260.31
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

room temp

SMILES string

CC(C(O)=O)c1ccc(cc1)C(=O)c2cccs2

InChI

1S/C14H12O3S/c1-9(14(16)17)10-4-6-11(7-5-10)13(15)12-3-2-8-18-12/h2-9H,1H3,(H,16,17)

InChI key

MDKGKXOCJGEUJW-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

Suprofen has been used to evaluate the combination of trifluoroacetic acid and isopropylamine (IPA) in supercritical fluid chromatography. It has also been used to evaluate immobilized chiral polysaccharide-based stationary phases in supercritical fluid chromatography.

Biochem/physiol Actions

Suprofen is a nonsteroidal anti-inflammatory drug that inhibits COX-1/2. Suprofen exhibits a potent anti-inflammatory, analgesic and antipyretic activities.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A S Nies
Agents and actions. Supplements, 24, 95-106 (1988-01-01)
All nonsteroidal anti-inflammatory drugs (NSAIDs) inhibit cyclooxygenase, and consequently renal functions dependent upon prostaglandin synthesis can be affected. Fortunately, renal function in normal individuals is relatively independent of the PG system, and thus the NSAIDs don't usually produce any renal
E L Tolman et al.
Clinics in rheumatic diseases, 10(2), 353-368 (1984-08-01)
Suprofen is a potent, peripherally-acting, non-narcotic analgesic agent. The mechanism of action of the compound involves inhibition of prostaglandin biosynthesis and, perhaps, direct antagonism of the peripheral, pain inducing actions of prostaglandins, bradykinin and other pain mediators. Suprofen at a
Pharmaceutical-enantiomers resolution using immobilized polysaccharide-based chiral stationary phases in supercritical fluid chromatography
De Klerck
Journal of Chromatography A (2014)
C J Niemegeers et al.
Arzneimittel-Forschung, 25(10), 1505-1509 (1975-10-01)
A new standardized acetic acid-induced writhing test in rats is described in detail and its methodology is discussed briefly. The described method has proved to be useful for evaluating the anti-writhing activity of narcotic analgesics, non-narcotic anti-inflammatory compounds and narcotic-antagonists
Malin Darnell et al.
Chemical research in toxicology, 28(5), 886-896 (2015-03-25)
Xenobiotic carboxylic acids may be metabolized to oxidative metabolites, acyl glucuronides, and/or S-acyl-CoA thioesters (CoA conjugates) in vitro, e.g., in hepatocytes, and in vivo. These metabolites can potentially be reactive species and bind covalently to tissue proteins and are generally

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service