Skip to Content
Merck
All Photos(3)

Key Documents

N27209

Sigma-Aldrich

4-Nitrothiophenol

technical grade, 80%

Synonym(s):

p-Nitrophenyl mercaptan, pNTP, 4-Nitrobenzenethiol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H4SH
CAS Number:
Molecular Weight:
155.17
Beilstein:
606924
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Assay

80%

form

solid

mp

72-77 °C (lit.)

storage temp.

2-8°C

SMILES string

[O-][N+](=O)c1ccc(S)cc1

InChI

1S/C6H5NO2S/c8-7(9)5-1-3-6(10)4-2-5/h1-4,10H

InChI key

AXBVSRMHOPMXBA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4-Nitrothiophenol has been used in the synthesis of dual emission fluorescent probe for the differential sensing of of glutathione (GSH) and cysteine/homocysteine (Cys/Hcy). pNTP can also be used to synthesize diaryl thioethers via copper-catalyzed C-S coupling reaction.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qiaofeng Yao et al.
Nature communications, 8(1), 1555-1555 (2017-11-18)
Precise control of alloying sites has long been a challenging pursuit, yet little has been achieved for the atomic-level manipulation of metallic nanomaterials. Here we describe utilization of a surface motif exchange (SME) reaction to selectively replace the surface motifs
Ze-Yuan Dong et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 12(13), 3575-3579 (2006-02-24)
To elucidate the relationships between molecular recognition and catalytic ability, we chose three assay systems using three different thiol substrates, glutathione (GSH), 3-carboxyl-4-nitrobenzenethiol (CNBSH), and 4-nitrobenzenethiol (NBSH), to investigate the glutathione peroxidase (GPx) activities of 2,2'-ditellurobis(2-deoxy-beta-cyclodextrin) (2-TeCD) in the presence
B O Skadtchenko et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 57A(5), 1009-1016 (2001-05-26)
The interpretation of the surface-enhanced Raman scattering (SERS) spectra of p-nitrothiophenol (p-NTP) is reported. SERS spectra were obtained by vacuum evaporation and casting of p-NTP onto silver island films, and also from colloidal silver solutions. The vibrational spectra of the
M Yamamoto et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 24(3), 247-250 (1986-03-01)
A simple and sensitive method for the determination of N-nitrosomethylurea (NMU) in biological fluids was developed. The principle is based on the formation of 4-nitrothiophenol methyl thioether (NTP-Me) from 4-nitrothiophenol (NTP) and diazomethane formed from MNU by adding alkali. After
Camiel H van Hoorn et al.
Applied spectroscopy, 71(7), 1551-1559 (2017-07-01)
A fast high-resolution screening method for reactive surfaces is presented. Atomic force microscopy (AFM) and surface-enhanced Raman spectroscopy (SERS) are combined in one method in order to be able to obtain both morphological and chemical information about processes at a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service