Skip to Content
Merck
All Photos(1)

Documents

C8249

Sigma-Aldrich

Cardamonin

≥98% (HPLC)

Synonym(s):

(2E)-1-(2,4-Dihydroxy-6-methoxyphenyl)-3-phenyl-2-propen-1-one, (8CI); Alpinetin chalcone, (E)-2′,4′-Dihydroxy-6′-methoxy-chalcone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H14O4
CAS Number:
Molecular Weight:
270.28
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

light yellow to dark brown

solubility

DMSO: ≥20 mg/mL

storage temp.

room temp

SMILES string

O=C(/C=C/C1=CC=CC=C1)C2=C(O)C=C(O)C=C2OC

InChI

1S/C16H14O4/c1-20-15-10-12(17)9-14(19)16(15)13(18)8-7-11-5-3-2-4-6-11/h2-10,17,19H,1H3/b8-7+

InChI key

NYSZJNUIVUBQMM-BQYQJAHWSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Cardamonin has been used as a glycogen synthase kinase 3 (GSK3) inhibitor to study its effects on β-catenin loss and gain-of-function in human preimplantation embryos.
Cardamonin has been used as a standard in high performance liquid chromatography (HPLC) to analyze phenolic compounds in acacia honey samples.

Biochem/physiol Actions

Cardamonin is a calchone from Aplinia species (zingiberaceous plant species). Cardamonin inhibits pigmentation in melanocytes through suppression of Wnt/b-catenin signaling pathway. Cardamonin suppressed Wnt/b-catenin signaling by a mechanism involving proteasome-mediated degradation of b-catenin (GSK-3b-independent pathway). Research has shown that Cardamonin possesses anti-inflammatory activity via suppression of NF-kB nuclear translocation and Ik-Ba phosphorylation.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bokyung Sung et al.
PloS one, 8(5), e64118-e64118 (2013-05-22)
Bone loss/resorption or osteoporosis is a disease that is accelerated with aging and age-associated chronic diseases such as cancer. Bone loss has been linked with human multiple myeloma, breast cancer, and prostate cancer and is usually treated with bisphosphonates, and
Peiguang Niu et al.
Oncology letters, 15(3), 3991-3997 (2018-02-20)
The mammalian target of rapamycin (mTOR) is well-known as a promising therapeutic target in various cancer cells. mTOR activation decreases the sensitivity of ovarian cancer to cisplatin. Cardamonin inhibits the proliferation of various cancer cells by mTOR suppression. The present
Jianqing Jiang et al.
Journal of microbiology and biotechnology, 31(6), 794-802 (2021-05-25)
In this study we investigated the role and mechanism of cardamonin on IL-1β induced injury in OA. CHON-001 cells were treated with cardamonin and IL-1β and transfected with silencing nuclear factor erythroid 2-related factor 2 (siNrf2). Cell viability was detected
Phenolics and abscisic acid identified in acacia honey comparing different SPE cartridges coupled with HPLC-PDA
Sun C, et al.
Journal of food composition and analysis, 53 (2016)
Chih-Chien Wang et al.
Medicina (Kaunas, Lithuania), 57(9) (2021-09-29)
Background and Objectives: Gouty arthritis is an acute inflammatory response caused by the precipitation of monosodium urate (MSU) crystals in joints. The triggering of MSU leads to increased production of inflammatory cytokines, such as interleukin-1β, which in turn lead to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service