Skip to Content
Merck
All Photos(3)

Key Documents

S9754

Sigma-Aldrich

Sulfatase from abalone entrails

Type VIII, lyophilized powder, 20-40 units/mg solid

Synonym(s):

Aryl-sulfatase, Aryl-sulfate sulfohydrolase, Phenolsulfatase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

biological source

abalone entrails

Quality Level

type

Type VIII

form

lyophilized powder

specific activity

20-40 units/mg solid

mol wt

~43 kDa by gel filtration

foreign activity

β-Glucuronidase ≤10.0 Sigma units/mg solid

storage temp.

−20°C

Application

Sulfatase from abalone entrails has been used in a study to determine that human p-selectin glycoprotein ligand-1 interacts with the skin-associated chemokine CCL27 via sulfated tyrosines at the PSGL-1 amino terminus. Sulfatase from abalone entrails has also been used in a study to investigate HPLC of sulfate and glutathione conjugates from hamster embryo fibroblasts.
Sulfatase from abalone entrails has been used to digest the conjugated forms into free forms for the analysis of tissue green tea polyphenols (GTPs). It has also been used for the detection of free, glucuronidated or total form of GTPs.

Biochem/physiol Actions

Sulfatases hydrolyze the sulfate esters of complex macromolecules, like glycosaminoglycans and sulfatides. It helps to modulate the sulfation states that determine the action of several physiological molecules.

Unit Definition

One unit will hydrolyze 1.0 μmole of p-nitrocatechol sulfate per hr at pH 5.0 at 37 °C (30 min assay).

substrate

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Quercetin increased bioavailability and decreased methylation of green tea polyphenols in vitro and in vivo
Wang P, et al.
Food, 3(6), 635-642 (2012)
Use of a Phage Display Antibody to Measure the Enzymatic Activity of the Sulfs
Uchimura K, et al.
Test, 480, 51-64 (2010)
B A Merrick et al.
Carcinogenesis, 6(9), 1303-1307 (1985-09-01)
Chromatography of benzo[a]pyrene (BaP) sulfate, glucuronide and glutathione (GSH) conjugate standards were examined by h.p.l.c. on a C8 column as modified by various organic acids and solvents. Sulfate and glucuronide standards were positional isomers derived from BaP-1,3,6,7,9 phenols and BaP-GSH
Sulfatases: structure, mechanism, biological activity, inhibition, and synthetic utility
Hanson SR, et al.
Angewandte Chemie (International ed. in English), 43(43), 5736-5763 (2004)
Green Tea Polyphenols and Metabolites in Prostatectomy Tissue: Implications for Cancer Prevention
Wang P, et al.
Cancer Prevention Research (Philadelphia, Pa.), 3(8), 985-993 (2010)

Protocols

Enzymatic Assay of Sulfatase (EC 3.1.6.1.)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service