RAB0676
Human MET / Hepatocyte Growth Factor Receptor ELISA Kit
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116158
NACRES:
NA.32
Recommended Products
species reactivity
human
packaging
kit of 96 wells (12 strips x 8 wells)
technique(s)
ELISA: suitable
input
sample type serum
sample type plasma
sample type cell culture supernatant(s)
assay range
inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 60 pg/mL
standard curve range: 61.4-15000 pg/mL
detection method
colorimetric
shipped in
wet ice
storage temp.
−20°C
Gene Information
human ... MET(4233)
General description
The antibody pair provided in this kit recognizes human Hepatocyte Growth Factor Receptor.
Application
For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Other Notes
A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.
Please type the word sample in the text box provided for lot number.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Met. Corr. 1
Storage Class Code
8A - Combustible corrosive hazardous materials
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Xinhong Wang et al.
Cytokine, 99, 275-280 (2017-08-24)
To evaluate the influence of overexpression HGF on the healing of traumatic ulcer of oral mucosa of mice. Mice were divided into two groups: wild type C57BL6(WT) and HGF high expression transgenic (HGF-Tg) mice. Traumatic ulcer of all mice were
Omidreza Firuzi et al.
Cancers, 11(5) (2019-05-11)
Pancreatic stellate cells (PSCs) are a key component of tumor microenvironment in pancreatic ductal adenocarcinoma (PDAC) and contribute to drug resistance. c-MET receptor tyrosine kinase activation plays an important role in tumorigenesis in different cancers including PDAC. In this study
Yan-Tao Fu et al.
American journal of translational research, 9(6), 3138-3147 (2017-07-04)
Tyrosine kinase receptors such as c-Met and its ligands are interesting therapeutic targets that have been reported to be involved in the progression of several types of cancers. Histone deacetylase inhibitor, valproic acid (VPA) is one such compound with promising
Pauline Brige et al.
American journal of physiology. Gastrointestinal and liver physiology, 315(1), G117-G125 (2018-02-23)
To reduce the morbidity and mortality risk for the donor in living donor liver transplantation (LDLT), we previously identified 20% left portal vein (LPV) stenosis as an effective preconditioning method to induce cell proliferation in the contralateral lobe without downstream
Ji-Dong Chen et al.
Asian Pacific journal of cancer prevention : APJCP, 15(11), 4545-4548 (2014-06-28)
To investigate the changes of serum vascular endothelial growth factor (VEGF), soluble interleukin-2 receptor (SIL-2R) and hepatocyte growth factor (HGF) contents in patients with primary hepatocellular carcinoma (HCC) before and after percutaneous microwave coagulation therapy (PMCT) and determine their clinical
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service