RAB0096
Human C-Reactive Protein ELISA Kit
for serum, plasma, cell culture supernatants and urine
Synonym(s):
C-reactive protein, CRP
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116158
NACRES:
NA.32
Recommended Products
species reactivity
human
packaging
kit of 96 wells (12 strips x 8 wells)
technique(s)
ELISA: suitable
capture ELISA: suitable
input
sample type cell culture supernatant(s)
sample type urine
sample type serum
sample type plasma
assay range
inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 34 pg/mL
standard curve range: 34.29-25000 pg/mL
detection method
colorimetric
shipped in
wet ice
storage temp.
−20°C
Gene Information
human ... CRP(1401)
Related Categories
General description
The Human CRP (C Reactive Protein) ELISA (Enzyme- Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human CRP in serum, plasma, cell culture supernatants and urine.
Immunogen
Recombinant Human CRP
Application
Human C-Reactive Protein ELISA Kit has been used to measure C-reactive protein (CRP) levels in serum.
Biochem/physiol Actions
C-reactive protein (CRP) is part of the pentraxin protein family and is synthesized in the liver. It is a marker of systemic inflammation and may also have a role in atherosclerosis, stroke and myocardial infarction. Levels of CRP increase during tissue damage and trauma. The protein acts as an opsonin and binds to phospholipids on tumor cell membranes. This may lead to lysis of tumor cells. CRP is also a biomarker of age-related macular degeneration (AMD).
Other Notes
A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.
Please type the word sample in the text box provided for lot number.
Kit Components Also Available Separately
Product No.
Description
SDS
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Met. Corr. 1
Storage Class Code
8A - Combustible corrosive hazardous materials
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Ying Liu et al.
Experimental and therapeutic medicine, 15(1), 585-591 (2018-02-02)
The objective of the present study was to investigate the role of Toll-like receptor (TLR)-9 in B lymphocyte stimulating factor (BLyS)-induced systemic lupus erythematosus (SLE) in mice. The anti-double stranded (ds)DNA antibody titer, levels of complement proteins (C3 and C4)
Adsorbing/dissolving Lyoprotectant Matrix Technology for Non-cryogenic Storage of Archival Human Sera
Solivio MJ, et al.
Scientific Reports, 6, 24186-24186 (2016)
Biomarker and shear stress in secondary pediatric pulmonary hypertension
Pektacs A, et al.
Turkish Journal of Medical Sciences, 47(6), 1854-1860 (2017)
Azza M Elamir et al.
Biomedical reports, 15(5), 93-93 (2021-10-12)
The aim of the present study was to detect the serum levels of long non-coding RNA (lncRNA) growth arrest-specific 5 (GAS5) in patients with rheumatoid arthritis (RA) and healthy controls, and determine the association between the rs2067079, rs6790, and rs17359906
Prognostic impact of C-reactive protein/albumin ratio on the overall survival of patients with advanced nonsmall cell lung cancers receiving palliative chemotherapy
Young W
Medicine (2017)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service