Skip to Content
Merck

Skip To

RABTMB3

ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.32
UNSPSC Code:
41116158
Storage temp.:
−20°C
Shipped in:
wet ice
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


technique(s)

ELISA: suitable

shipped in

wet ice

storage temp.

−20°C

Other Notes

Intended for use with ELISA kits.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
RABTMB1ELLURELLUF0100
description

-

description

-

description

Western blotting, dot/slot blotting

description

Western blotting, dot/slot blotting

technique(s)

ELISA: suitable

technique(s)

-

technique(s)

ELISA: suitable

technique(s)

ELISA: suitable

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

storage temp.

−20°C

storage temp.

−20°C

storage temp.

-

storage temp.

2-8°C


Storage Class

12 - Non Combustible Liquids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters