RAB0510
Mouse VEGF ELISA Kit
for cell and tissue lysates
Synonym(s):
VEGF-A
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116158
NACRES:
NA.32
Recommended Products
species reactivity
mouse
packaging
kit of 96 wells (12 strips x 8 wells)
technique(s)
ELISA: suitable
capture ELISA: suitable
input
sample type cell lysate
sample type tissue lysate
assay range
inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 2 pg/mL
standard curve range: 4.1-1000 pg/mL
detection method
colorimetric
shipped in
wet ice
storage temp.
−20°C
Gene Information
mouse ... Vegfa(22339)
Related Categories
General description
The Mouse VEGF ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of mouse VEGF cell lysate and tissue lysate.
Immunogen
Recombinant Mouse VEGF-A
Application
For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Mouse VEGF ELISA Kit has been used to determine the concentration of VEGF expression in samples using ELISA.
Biochem/physiol Actions
Vascular endothelial growth factor (VEGF) is a critical factor in regulating angiogenesis in both physiological and pathological conditions. Transforming growth factor-α (TGF-α), TGF-β and platelet derived growth factor can potentially activate VEGF activity. VEGF mediates its activity through tyrosine kinase receptors, vascular endothelial growth factor receptor-1 (VEGFR-1) and VEGFR-2. VEGF produced by tumour cells and macrophages, plays a key role in cancer development by promoting endothelial cells proliferation and increasing vascular permeability. Enhanced expression of VEGF plays a key role in urologic neoplasm like bladder cancer, renal cell carcinoma, and cervical cancer. Polymorphism in VEGF gene increases the disease susceptibility. Blocking of VEGF mediated signalling pathway is a potential therapeutic strategy for cancer patients.
Other Notes
A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.
Please type the word sample in the text box provided for lot number.
Kit Components Also Available Separately
Product No.
Description
SDS
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Met. Corr. 1
Storage Class Code
8B - Non-combustible corrosive hazardous materials
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Relationship of common vascular endothelial growth factor polymorphisms and haplotypes with the risk of cervical cancer in Tunisians
Zidi S, et al.
Cytokine, 74(1), 108-112 (2015)
Stimulation of angiogenesis using single-pulse low-pressure shock wave treatment
Sundaram S, et al.
Journal of Molecular Medicine, 96(11), 1177-1187 (2018)
Parthenolide inhibits tumor-promoting effects of nicotine in lung cancer by inducing P53-dependent apoptosis and inhibiting VEGF expression
Talib WH, et al.
Biomedicine and Pharmacotherapy, 107, 1488-1495 (2018)
Fu-Li Xiang et al.
Circulation. Heart failure, 7(5), 831-842 (2014-08-12)
The adult epicardium is a potential source of cardiac progenitors after myocardial infarction (MI). We tested the hypothesis that cardiomyocyte-specific overexpression of membrane-associated human stem cell factor (hSCF) enhances epicardial activation, epicardium-derived cells (EPDCs) production, and myocardial arteriogenesis post MI.
Combination of metformin and curcumin targets breast cancer in mice by angiogenesis inhibition, immune system modulation and induction of p53 independent apoptosis
Falah RR, et al.
Therapeutic Advances in Medical Oncology, 9(4), 235-252 (2017)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service