Skip to Content
Merck
All Photos(1)

Key Documents

RAB0509

Sigma-Aldrich

Mouse VEGF ELISA Kit

for serum, plasma, cell culture supernatant

Synonym(s):

VEGF-A, Vascular endothelial growth factor A 

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.84
Pricing and availability is not currently available.

species reactivity

mouse

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
capture ELISA: suitable

input

sample type cell culture supernatant(s)
sample type serum
sample type plasma

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 2 pg/mL
standard curve range: 4.1-1000 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

mouse ... Vegfa(22339)

General description

Vascular endothelial growth factor (VEGF) is produced by thymus epithelial cells in adults. It is located on human chromosome 6p21.1[1] The Mouse VEGF ELISA (enzyme-linked immunosorbent assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of mouse VEGF in serum, plasma and cell culture supernatants.

Immunogen

Recombinant Mouse VEGF-A

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Mouse VEGF ELISA Kit has been used in biochemical analysis.[2][3]

Biochem/physiol Actions

Vascular endothelial growth factor (VEGF) supports vascular homeostasis.[4] Vascular endothelial growth factor-A (VEGF-A) modulates angiogenesis, vascular permeability and growth. It plays a vital role in the progression of the vasculature. Deficiency of VEGF-A leads to endothelial dysfunction, hypertension, ischemia and blood clotting.[5]

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABELADAELISA 1X Assay/Sample Diluent Buffer A (Item D1)SDS

  • RABELADBELISA 5X Assay/Sample Diluent Buffer B (Item E1)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

REGULATION OF VASCULAR ENDOTHELIAL GROWTH FACTOR (VEGF) PRODUCTION BY MOUSE THYMIC EPITHELIAL CELL LINES.
Rutto K V, et al.
Tsitologiia, 58(6), 436-443 (2016)
Jia-Wen Zhan et al.
PloS one, 15(6), e0234747-e0234747 (2020-06-26)
The vascular buds in the vertebral endplate (VEP) are the structural foundation of nutrient exchange in the intervertebral disc (IVD). VEGF is closely related to angiogenesis in the endplate and intervertebral disc degeneration (IDD). To investigate the effects of static
In situ functionalized nanobiocomposites dressings of bamboo cellulose nanocrystals and silver nanoparticles for accelerated wound healing
Singla R, et al.
Carbohydrate Polymers, 155(1), 152-162 (2017)
Doxycycline modulates VEGF-A expression: Failure of doxycycline-inducible lentivirus shRNA vector to knockdown VEGF-A expression in transgenic mice
Merentie M, et al.
Testing, 13(1), e0190981-e0190981 (2018)
Hsiao-Ying Chou et al.
Polymers, 12(9) (2020-09-24)
Polymer-based hydrogels demonstrate superior performance when used as wound dressing. An ideal dressing should possess an active healing function, absorb wound exudates, and provide a moist interface on the wound for rapid injury repair and the prevention of pain and

Questions

  1. Can you confirm that your VEGF-A Mouse ELISA Kit, RAB0509-1KT, is capable of detecting all three isoforms - VEGF120, VEGF164, and VEGF188?

    1 answer
    1. The antibodies provided in RAB0509-1KT are capable of specifically detecting VEGF120 and VEGF164, but not VEGF188.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service