RAB0382
Human NGF R ELISA Kit
for serum, plasma, cell culture supernatant and urine
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
41116158
NACRES:
NA.32
Recommended Products
species reactivity
human
packaging
kit of 96 wells (12 strips x 8 wells)
technique(s)
ELISA: suitable
capture ELISA: suitable
input
sample type cell culture supernatant(s)
sample type urine
sample type serum
sample type plasma
assay range
inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 80 pg/mL
standard curve range: 0.082-20 ng/mL
detection method
colorimetric
shipped in
wet ice
storage temp.
−20°C
Gene Information
human ... NGFR(4804)
General description
The Human NGFR (Nerve Growth Factor Receptor) ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme- linked immunosorbent assay for the quantitative measurement of human NGFR in serum, plasma, cell culture supernatants and urine.
Immunogen
Recombinant Human NGFR
Application
For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Other Notes
A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.
Please type the word sample in the text box provided for lot number.
Kit Components Also Available Separately
Product No.
Description
SDS
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Met. Corr. 1
Storage Class Code
8A - Combustible corrosive hazardous materials
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Takuya Noda et al.
Chemical senses, 44(5), 327-338 (2019-04-17)
Post-upper respiratory tract infection related olfactory dysfunction typically occurs due to neural damage after an upper respiratory tract infection associated with a common cold or influenza. At present, Tokishakuyakusan, a Japanese traditional Kampo medicine, has been found to be effective
Alireza Majdi et al.
Frontiers in aging neuroscience, 10, 194-194 (2018-08-01)
Here, we tested the claim that nicotine attenuates the signs of brain dysfunction in the model of brain aging induced by D-galactose (DGal) in mice. We administered nicotine at doses of 0.1, 0.5 and 1 mg/kg by the subcutaneous (s.c.)
Yusuke Sakurai et al.
Pain, 160(4), 895-907 (2018-12-27)
Most advanced knee osteoarthritis (OA) patients experience chronic pain resistant to cyclooxygenase (COX) inhibitors. However, the cells and molecules involved in this advanced OA pain remain poorly understood. In this study, we developed a rat model of advanced knee OA
Metin Uz et al.
Acta biomaterialia, 56, 141-152 (2016-10-04)
In this study, a poly(lactic acid) (PLLA) porous film with longitudinal surface micropatterns was fabricated by a dry phase inversion technique to be used as potential conduit material for peripheral nerve regeneration applications. The presence of a nerve growth factor
Angélica Carbajal-Ramírez et al.
BMC neurology, 17(1), 161-161 (2017-08-27)
Neuropathy is one of the major complications of type 2 diabetes mellitus. Our first aim was to determine the clinical characteristics of a population of diabetic patients with different types of neuropathy. Our next goal was to characterize the cytokine
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service