Skip to Content
Merck

HS120879

Slide staining tray with lid

to hold, 18 x slides 76x26 mm

Synonym(s):

Stain tray

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.13
UNSPSC Code:
41122800
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

black plastic

sterility

non-sterile

feature

to hold 18 x slides 76x26 mm

packaging

pack of 1 ea

manufacturer/tradename

Heathrow Scientific 120879

trays L × W × H

16.1 in. (41 cm) × 8.9 in. (22.5 cm) × 1.6 in. (4 cm)

General description

Ideal for Clinical and Research laboratory staining
Tray holds 18 standard microscope slides (76 x 26 mm) securely in place for staining, rinsing and drying applications

Contains one each of the following components:
1 Black base/tray
1 Black cover
2 drain plugs

Application

For Immunohistochemistry

Features and Benefits

Robust design
· Durably constructed from solvent resistant polypropylene blend
· Non-skid Silicone feet keep tray in place
· Convenient to use
· Two ways to empty stain liquid
Pour spouts on each corner
Drain hole with plug
· Equipped with handle cut outs for easy handling

Versatile for multiple testing formats
· Black lid included for light sensitive applications
· Humidity retention wells for specialized staining needs




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library