Skip to Content
Merck
All Photos(2)

Key Documents

RAB0123

Sigma-Aldrich

Human SDF 1 α ELISA Kit

for plasma, cell culture supernatant and urine

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32
Pricing and availability is not currently available.

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
capture ELISA: suitable

input

sample type cell culture supernatant
sample type urine
sample type plasma

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 80 pg/mL
standard curve range: 93.75-6000 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... CXCL12(6387)

General description

The Human SDF-1 α ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human SDF-1 α in plasma, cell culture supernatants and urine.

Immunogen

Recombinant Human SDF-1 α

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABELADAELISA 1X Assay/Sample Diluent Buffer A (Item D1)SDS

  • RABELADBELISA 5X Assay/Sample Diluent Buffer B (Item E1)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The chemokine SDF-1 is a chemoattractant for human CD34+ hematopoietic progenitor cells and provides a new mechanism to explain the mobilization of CD34+ progenitors to peripheral blood.
Aiuti A
The Journal of Experimental Medicine, 185(1), 111-120 (1997)
Wnt/?-catenin pathway is required for epithelial to mesenchymal transition in CXCL12 over expressed breast cancer cells.
Shan S et.al.
International Journal of Clinical and Experimental Pathology, 8(10), 12357-12367 (2015)
Gene-environment interaction effects on the development of immune responses in the 1st year of life.
Hoffjan S
American Journal of Human Genetics, 76(4), 696-704 (2005)
Endomyocardial expression of SDF-1 predicts mortality in patients with suspected myocarditis.
Zuern CS
Clinical Research in Cardiology : Official Journal of the German Cardiac Society, 104(12), 1033-1043 (2015)
The diagnostic and prognostic value of serum CXCL12 levels in patients with ischemic stroke.
Duan XX
Neurological Sciences, 36(12), 2227-2234 (2015)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service