RAB0273
Human IL-1 β ELISA Kit
for serum, plasma, cell culture supernatant and urine
Synonym(s):
Il-1 beta, Interleukin-1 beta
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
41116158
NACRES:
NA.84
Recommended Products
species reactivity
human
packaging
kit of 96 wells (12 strips x 8 wells)
technique(s)
ELISA: suitable
capture ELISA: suitable
input
sample type urine
sample type serum
sample type plasma
sample type cell culture supernatant(s)
assay range
inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 0.3 pg/mL
standard curve range: 0.48-100 pg/mL
detection method
colorimetric
shipped in
wet ice
storage temp.
−20°C
Gene Information
human ... IL1B(3553)
General description
The Human IL-1 β ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human IL-1 β in serum, plasma, cell culture supernatants and urine.
Immunogen
Recombinant Human IL-1β
Application
For research use only. Not for use in diagnostic procedures.
Human IL-1 β ELISA kit has been used to measure the concentration of interleukin 1β (IL-1 β) in cell culture supernatants.
Human IL-1 β ELISA kit has been used to measure the concentration of interleukin 1β (IL-1 β) in cell culture supernatants.
Biochem/physiol Actions
Interleukin 1β (IL-1 β) is a proinflammatory cytokine, which is implicated in induction of various acute and chronic inflammation. Hence, IL1B can be considered as a potential therapeutic target for disease associated with fibrosis and tissue remodeling. IL-1β protein increases the production of endogenous 92kDa gelatinase (matrix metalloproteinases 9 (MMP-9)) and tissue inhibitors of MMP (TIMP -1). Elevated concentration of IL-1β in synovial fluid contributes to the pathogenesis of synovitis and degenerative changes of the cartilaginous tissue and bone of the temporomandibular joint. IL-1β brings neutrophil and macrophage to the site of infection. Mutations in IL-1β gene are linked with renal manifestations and renal sequelae in Henoch-Schönlein purpura (HSP).
Other Notes
A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.
Please type the word sample in the text box provided for lot number.
Kit Components Also Available Separately
Product No.
Description
SDS
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Met. Corr. 1
Storage Class Code
8A - Combustible corrosive hazardous materials
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Jie Liu et al.
The Journal of international medical research, 49(3), 300060521996564-300060521996564 (2021-03-27)
Porphyromonas gingivalis (Pg) plays a critical role in the occurrence and development of atherosclerosis. Lipopolysaccharide from Pg (Pg-LPS) could lead to pyroptosis of vascular smooth muscle cells (VSMCs) and induce instability of atherosclerotic plaque. Therefore, pyroptosis of VSMCs could promote
Proinflammatory cytokines detectable in synovial fluids from patients with temporomandibular disorders.
Takahashi T
Oral Surgery, Oral Medicine, Oral Pathology, Oral Radiology, and Endodontics, 85(2), 135-141 (1998)
Zetao Ma et al.
Journal of orthopaedic surgery and research, 15(1), 284-284 (2020-07-30)
Inflammation and apoptosis of chondrocytes are the pathological bases of osteoarthritis. Autophagy could alleviate the symptoms of inflammation and apoptosis. Previous study has shown that BCL2/adenovirus E1B 19 kDa protein-interacting protein 3 (BNIP3) can induce the occurrence and development of autophagy.
Zhao Gao et al.
BMC ophthalmology, 20(1), 134-134 (2020-04-08)
Inflammation of RPE cells led to different kinds of eye diseases and affected the normal function of the retina. Furthermore, higher levels of ROCK1 and ROCK2 induced injury of endothelial cells and many inflammatory diseases of the eyes. Ripasudil, which
Interleukin 1 beta (IL1?) rs16944 genetic variant as a genetic marker of severe renal manifestations and renal sequelae in Henoch-Schonlein purpura.
Lopez-Mejias R
Clinical and Experimental Rheumatology, 34(3 Suppl 97), S84-S88 (2016)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service