Skip to Content
Merck
All Photos(4)

Key Documents

R6750

Sigma-Aldrich

Ribonucleic acid from baker′s yeast (S. cerevisiae)

Synonym(s):

RNA

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41106305
NACRES:
NA.51
Pricing and availability is not currently available.

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Ribonucleic Acid (RNA) from baker’s yeast (S. cerevisiae) is the substrate for RNase enzyme. The isolation of RNA from yeast is a complicated process and involves heating and freezing cycles of cells in the presence of phenyl and detergents (SDS).[1]

Application

Ribonucleic Acid (RNA) from baker’s yeast (S. cerevisiae) is suitable for applications such as northern blot hybridization, reverse transcriptase-polymerase chain reaction (RTPCR), and cDNA construction.[1]

Preparation Note

Prepared by a modification of Crestfield, A.M., et al., J. Biol. Chem., 216, 185 (1955).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kazuya Shirato et al.
Virology journal, 11, 139-139 (2014-08-12)
The first documented case of Middle East Respiratory Syndrome coronavirus (MERS-CoV) occurred in 2012, and outbreaks have continued ever since, mainly in Saudi Arabia. MERS-CoV is primarily diagnosed using a real-time RT-PCR assay, with at least two different genomic targets
Michael R Green et al.
Cold Spring Harbor protocols, 2021(12) (2021-12-03)
Isolation of RNA from yeast is complicated by the need to first break the thick, rigid cell wall. The protocol provided here uses a cycle of heating and freezing of cells in the presence of phenol and the detergent sodium
Radha Duttagupta et al.
PloS one, 7(2), e31241-e31241 (2012-02-24)
Inflammatory Bowel Disease--comprised of Crohn's Disease and Ulcerative Colitis (UC)--is a complex, multi-factorial inflammatory disorder of the gastrointestinal tract. In this study we have explored the utility of naturally occurring circulating miRNAs as potential blood-based biomarkers for non-invasive prediction of
Stephanie C Bannister et al.
Biology of reproduction, 81(1), 165-176 (2009-04-10)
MicroRNAs are a highly conserved class of small RNAs that function in a sequence-specific manner to posttranscriptionally regulate gene expression. Tissue-specific miRNA expression studies have discovered numerous functions for miRNAs in various aspects of embryogenesis, but a role for miRNAs
Douglas A Weigent
Cellular immunology, 282(2), 71-78 (2013-06-19)
In the present study, we show that higher levels of lymphocyte GH are expressed in spleen cells from aging animals compared to young animals. Further, leukocytes from primary and secondary immune tissues and splenic T and B cells from aging

Protocols

This procedure may be used for determination of Ribonuclease A (RNase A) activity.

This procedure may be used for determination of Ribonuclease A (RNase A) activity.

This procedure may be used for determination of Ribonuclease A (RNase A) activity.

This procedure may be used for determination of Ribonuclease A (RNase A) activity.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service