Skip to Content
Merck
All Photos(1)

Key Documents

10165948001

Roche

RNA, MS2

from bacteriophage MS2

Synonym(s):

RNA, ribonucleic acid

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106311

biological source

bacteriophage MS2

Quality Level

form

solution

mol wt

~1200 kDa

packaging

pkg of 500 μL (10 A260 units)

manufacturer/tradename

Roche

technique(s)

RNA extraction: suitable

color

colorless

solubility

water: miscible

storage temp.

−20°C (−15°C to −25°C)

Related Categories

General description

RNA, from bacteriophage MS2 is composed of 3569 nucleotides. MS2 bacteriophage acts as a model system in studying molecular biology processes, such as viral RNA replication, process of translation and physiology of infected cells. MS2 RNA codes for three viral polypeptides namely, protein A, coat protein and RNA replicase complex. Protein A and coat protein make up the structure of MS2 virus.

Application

RNA, MS2 has been used:
  • as a standard to assess RNA quantification methods
  • in RNA extraction
  • in single-tube, fluorescent PERT assay (STF-PERT) for accurate quantitation of different retrovirus types
  • for studies which use natural RNA in a in vivo and in vitro protein-synthesizing system, initiation, elongation, and termination of polypeptide synthesis.
  • for structural and functional studies.
  • to stabilize RNA during cDNA synthesis
  • to improve RNA yield during RNA extraction or isolation

Quality

Typical analysis: Free of protein and host nucleic acids according to current Quality Control procedures.

Sequence

Chain Length 3,569 nucleotides

Preparation Note

Working solution: Storage buffer: 10 mM Tris-HCl, 1 mM EDTA, pH 7.0.

Other Notes

For life science research only. Not for use in diagnostic procedures.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Monique I Andersson et al.
Wellcome open research, 5, 181-181 (2020-12-09)
Background: Laboratory diagnosis of SARS-CoV-2 infection (the cause of COVID-19) uses PCR to detect viral RNA (vRNA) in respiratory samples. SARS-CoV-2 RNA has also been detected in other sample types, but there is limited understanding of the clinical or laboratory
Kirstine Kirkegaard et al.
Journal of assisted reproduction and genetics, 37(9), 2199-2209 (2020-07-19)
miRNAs have been suggested as biomarkers of embryo viability; however, findings from preliminary studies are divergent. Furthermore, the presence of other types of small RNA molecules remains to be investigated. The purpose of this study was to perform a comprehensive
Dennis M Timmerman et al.
Cancers, 13(17) (2021-09-11)
MicroRNAs (miRNAs) are short, non-coding RNAs involved in translation regulation. Dysregulation has been identified in cancer cells. miRNAs can be secreted and detectable in body fluids; therefore, they are potential liquid-biopsy biomarkers. The miR-371a-3 cluster members are an example, monitoring
Petra Vychytilova-Faltejskova et al.
Carcinogenesis, 37(10), 941-950 (2016-08-04)
Early detection of colorectal cancer is the main prerequisite for successful treatment and reduction of mortality. Circulating microRNAs were previously identified as promising diagnostic, prognostic and predictive biomarkers. The purpose of this study was to identify serum microRNAs enabling early
Shalini Das Gupta et al.
Scientific reports, 10(1), 9012-9012 (2020-06-04)
Quantification of plasma microRNAs (miRNAs) as non-invasive disease biomarkers is subject to multiple technical variabilities. This study aimed to develop an optimized protocol for miRNA quantification from rodent plasma. We hypothesized that a fixed small RNA concentration input for reverse

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service