Skip to Content
Merck
All Photos(3)

Key Documents

P8375

Sigma-Aldrich

Peroxidase from horseradish

Type VI, essentially salt-free, lyophilized powder, ≥250 units/mg solid (using pyrogallol)

Synonym(s):

Detection Enzyme, HRP, Peroxidase, Donor:hydrogen-peroxide oxidoreductase, Horseradish peroxidase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
eCl@ss:
32160410
NACRES:
NA.54

type

Type VI

Quality Level

form

essentially salt-free, lyophilized powder

specific activity

≥250 units/mg solid (using pyrogallol)

mol wt

~44 kDa

solubility

0.1 M phosphate buffer: soluble 10 mg/mL, clear, orange to red (pH 6.0)

absorbance ratio

RZ 2.5-4.0

application(s)

diagnostic assay manufacturing

storage temp.

2-8°C

SMILES string

[O+H2]O[O-]

InChI

1S/H2O3/c1-3-2/h1-2H

InChI key

JSPLKZUTYZBBKA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Horseradish peroxidase is isolated from horseradish roots (Amoracia rusticana) and belongs to the ferroprotoporphyrin group of peroxidases. HRP is a single chain polypeptide containing four disulfide bridges. It is a glycoprotein containing 18% carbohydrate. The carbohydrate composition consists of galactose, arabinose, xylose, fucose, mannose, mannosamine, and galactosamine depending upon the specific isozyme. Its molecular weight (~44 kDa) includes the polypeptide chain (33,890 Daltons), hemin plus Ca2+ (~700 Daltons), and carbohydrate (~9,400 Daltons). At least seven isozymes of HRP exist. The isoelectric point for horseradish peroxidase isozymes ranges from 3.0 - 9.0.

Application

Horseradish peroxidase (HRP) is isolated from horseradish roots (Amoracia rusticana) and belongs to the ferroprotoporphyrin group of peroxidases.. It is used in biochemistry applications such as western blots, ELISA and Immunohistochemistry. Horseradish peroxidase is used to amplify a weak signal and increase detectability of a target molecule, such as a protein. Product P8375, type VI, is an essentially salt free lyophilized powder. It is commonly used to determine amounts of glucose and peroxides in solution. It has been used to study sensory input of cervical spinal cord neurons.
The enzyme has been used to develop a thermostable soybean peroxidase-based biosensor by cross-linking and electrically ′wiring′ the enzyme through a redox-conducting hydrogel to a glassy carbon electrode. It has also been used to assay the oxidation of low density lipoprotein.

Biochem/physiol Actions

HRP readily combines with hydrogen peroxide (H2O2) and the resultant [HRP-H2O2] complex can oxidize a wide variety of hydrogen donors. The optimal pH is 6.0-6.5 and the enzyme is most stable in the pH range of 5.0-9.0. HRP can be conjugated to antibodies by several different methods including glutaraldehyde, periodate oxidation, through disulfide bonds, and also via amino and thiol directed cross-linkers. It is smaller and more stable than the enzyme labels β-galactosidase and alkaline phosphatase and hence, it is the most desired label. Also, its glycosylation leads to lower non-specific binding.
When incubated with a substrate, horseradish peroxidase produces a coloured, fluorimetric, or luminescent derivative of the labeled molecule, allowing quantification. Horseradish peroxidase has been shown to slightly reduce the level of inhibition in a cydAB mutant. Known inhibitors are sodium azide, cyanide, L-cystine, dichromate, ethylenethiourea, hydroxylamine, sulfide, vanadate, p-aminobenzoic acid, and Cd2+, Co2+, Cu2+, Fe3+, Mn2+, Ni2+, and Pb2+ ions.

Unit Definition

One pyrogallol unit will form 1.0 mg purpurogallin from pyrogallol in 20 sec at pH 6.0 at 20 °C.

Analysis Note

The RZ (Reinheitszahl) is the absorbance ratio A403/A275 determined at 0.5-1.0 mg/ml in deionized water. It is a measure of hemin content, not enzymatic activity. Even preparations with high RZ may have low enzymatic activity.

Other Notes

View more information on peroxidase at www.sigma-aldrich.com/enzymeexplorer.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Thermostable Hydrogen Peroxide Sensor Based on "Wiring" of Soybean Peroxidase
Analytical Chemistry, 67 (23), 4247-4249 (1995)
Filip Mollerup et al.
PloS one, 14(5), e0216546-e0216546 (2019-05-16)
Copper radical alcohol oxidases belonging to auxiliary activity family 5, subfamily 2 (AA5_2) catalyze the oxidation of galactose and galactosides, as well as aliphatic alcohols. Despite their broad applied potential, so far very few AA5_2 members have been biochemically characterized.
Krishna Mohan Pathi et al.
Frontiers in plant science, 11, 543895-543895 (2020-11-17)
Biotic stresses caused by microbial pathogens impair crop yield and quality if not restricted by expensive and often ecologically problematic pesticides. For a sustainable agriculture of tomorrow, breeding or engineering of pathogen-resistant crop varieties is therefore a major cornerstone. Maize
Marilyn S Lee et al.
Synthetic and systems biotechnology, 5(3), 145-154 (2020-07-09)
Cell-free systems contain many proteins and metabolites required for complex functions such as transcription and translation or multi-step metabolic conversions. Research into expanding the delivery of these systems by drying or by embedding into other materials is enabling new applications
E Wieland et al.
Proceedings of the National Academy of Sciences of the United States of America, 90(13), 5929-5933 (1993-07-01)
Oxidative modification of low density lipoprotein is believed to be an important pathway by which the lipoprotein becomes atherogenic. The in vitro systems for oxidative modification of low density lipoprotein thus far described all appear to depend upon the presence

Articles

Discover our peroxidase from horseradish enzymes, products, substrates, and inhibitors for your ELISA, immunoassay, and protein application needs.

Discover our peroxidase from horseradish enzymes, products, substrates, and inhibitors for your ELISA, immunoassay, and protein application needs.

Discover our peroxidase from horseradish enzymes, products, substrates, and inhibitors for your ELISA, immunoassay, and protein application needs.

Discover our peroxidase from horseradish enzymes, products, substrates, and inhibitors for your ELISA, immunoassay, and protein application needs.

Protocols

Reinheitszahl (RZ) is the ratio of absorbance due to hemin (A403, Soret region) to absorbance due to protein (A275).

Reinheitszahl (RZ) is the ratio of absorbance due to hemin (A403, Soret region) to absorbance due to protein (A275).

Reinheitszahl (RZ) is the ratio of absorbance due to hemin (A403, Soret region) to absorbance due to protein (A275).

Reinheitszahl (RZ) is the ratio of absorbance due to hemin (A403, Soret region) to absorbance due to protein (A275).

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service