Skip to Content
Merck
All Photos(9)

Key Documents

C5896

Sigma-Aldrich

Choline Oxidase from Alcaligenes sp.

lyophilized powder, ≥10 units/mg solid

Synonym(s):

Choline: oxygen 1-oxidoreductase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

form

lyophilized powder

Quality Level

specific activity

≥10 units/mg solid

mol wt

~95 kDa

technique(s)

cell based assay: suitable

suitability

suitable for assay of choline
suitable for molecular biology

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Choline oxidase from Alcaligenes sp. has been used:
  • in a study to investigate a choline biosensor constructed with a chitinous membrane and its application in measuring cholinesterase inhibitory activities
  • has also been used in spectroscopic studies on the photoreaction of choline oxidase with covalently bound flavin.
  • as a component of phosphate buffer saline (PBS) buffer for measurement of choline acetyltransferase (ChAT) activity in brain tissue homogenates
  • to measure the choline generated by autotaxin (ATX) by ATX activity assay and phospholipase-D (PLD) by PLD activity assay

Biochem/physiol Actions

Choline oxidase catalyzes the four-electron oxidation of choline to glycine-betaine, with betaine-aldehyde as the intermediate and molecular oxygen as th eprimary electron acceptor. The enzyme can also accept betaine-aldehyde as a substrate. This allows the study of the reaction mechanism for the conversion of choline to the aldehyde intermediate, & of betaine-aldehyde to glycine-betaine. The enzyme is a flavoprotein with a molecular weight of approx. 72,000 Da (gel filtration) or 66,000 Da (SDS gel electrophoresis).

Physical properties

Isoelectric point:4.1 ± 0.1
Michaelis constants:2.84 x 10¯3M (Choline), 5.33 x 10¯3M (Betaine aldehyde)
Structure:One mol of FAD is covalently bound to mol of the enzyme
Inhibitors:p-Chloromercuribenzoate, Cu++, Co++, Hg++, Ag+
Optimum pH:8.0 – 8.5
Optimum temp:40 – 45°C
pH Stability:pH 7.0 – 9.0 (30°C, 2hr)
Thermal stability:Below 37°C (pH 7.5, 10min)

Unit Definition

One unit will form 1 μmole of H2O2 with oxidation of 1 μmole of choline to betaine aldehyde per min at pH 8.0 at 37 °C. Note: During the conversion of choline to betaine by choline oxidase, 2 μmoles of H2O2 are produced for every μmole of choline.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Ohta-Fukuyama et al.
Journal of biochemistry, 88(1), 197-203 (1980-07-01)
Choline oxidase from Alcaligenes sp. catalyzed the oxidation of choline and betaine aldehyde to betaine with concomitant consumption of oxygen and production of hydrogen peroxide. The values of Km for choline and betaine aldehyde were 0.87 and 6.2 mM, respectively.
S Pundir et al.
Analytical biochemistry, 427(1), 26-32 (2012-05-09)
A bienzymatic choline biosensor was constructed by coimmobilizing acetylcholinesterase (AChE) and choline oxidase (ChO) onto nanocomposite of carboxylated multiwalled carbon nanotubes (c-MWCNTs) and zirconium oxide nanoparticles (ZrO(2)NPs) electrodeposited on the surface of a glassy carbon electrode (GCE) and using it
Giovanni Gadda
Biochimica et biophysica acta, 1646(1-2), 112-118 (2003-03-15)
Choline oxidase catalyzes the four-electron oxidation of choline to glycine-betaine, with betaine-aldehyde as intermediate and molecular oxygen as primary electron acceptor. The enzyme is capable of accepting betaine-aldehyde as a substrate, allowing the investigation of the reaction mechanism for both
Shihua Hou et al.
Biosensors & bioelectronics, 33(1), 44-49 (2012-01-11)
A novel acetylcholinesterase (AChE)/choline oxidase (ChOx) bienzyme amperometric acetylcholine biosensor based on gold nanoparticles (AuNPs) and multi-walled carbon nanotubes (MWCNTs) has been successfully developed by self-assembly process in combination of sol-gel technique. A thiolated aqueous silica sol containing MWCNTs and
Shengyuan Deng et al.
Biosensors & bioelectronics, 26(11), 4552-4558 (2011-06-10)
A signal amplification system for electrochemiluminescence (ECL) of quantum dots (QDs) was developed by using electrochemically reduced graphene oxide (ERGO) to construct a nanobiosensing platform. Due to the structural defects of GO, the ECL emission of QDs coated on GO

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service