Skip to Content
Merck

77332

Peroxidase from horseradish

lyophilized, powder, ~150 U/mg

Synonym(s):

(2R,3S,6S,7R,8R)-3-(3-formamido-2-hydroxybenzamido)-8-hexyl-2,6-dimethyl-4,9-dioxo-1,5-dioxonan-7-yl 3-methylbutanoate, Antimycin A, Horseradish peroxidase

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
NACRES:
NA.54
UNSPSC Code:
12352204
EC Number:
232-668-6
MDL number:
EC Number:
eCl@ss:
32160410
Specific activity:
~150 U/mg
Biological source:
horseradish
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

horseradish

Quality Segment

form

powder

quality

lyophilized

specific activity

~150 U/mg

solubility

50 mM potassium phosphate buffer, pH 6.0: 5 mg/mL (0.1% BSA)

shipped in

wet ice

storage temp.

2-8°C

SMILES string

[O+H2]O[O-]

InChI

1S/H2O3/c1-3-2/h1-2H

InChI key

JSPLKZUTYZBBKA-UHFFFAOYSA-N

General description

Horseradish peroxidase (HRP) is a vital and a widely studied heme-containing enzyme, isolated from a hardy perennial herb, horse radish. It catalyzes the oxidation of various organic and inorganic compounds by using hydrogen peroxide as substrates. HRP is involved in indole-3-acetic acid metabolism, cross-linking of biological polymers and lignification. This enzyme in combination with indole-3-acetic acid or its derivatives acts as a potential therapeutic for targeted cancer therapies. Commercially HRP is used as a constituent of clinical diagnostic kits and is used in immunoassays.

Application

Horseradish peroxidase is used widely within biochemistry as a detection reagent in a variety of applications including ELISA, immunoblotting, and immunohistochemistry.
Peroxidase from horseradish has been used:
  • for isolation of N-Linked glycans
  • as a constituent of D-amino acid oxidase (DAAO) buffer
  • to decompose hydrogen peroxide (H2O2) into water to demonstrate the performance of the portable oxidative stress sensor (POSS)

Other Notes

1 U corresponds to the amount of enzyme which oxidizes 1 μmol ABTS (Cat. No. 11557) per minute at pH 6.0 and 25°C
Use of chemically modified peroxidase in benzene; Cross-linking of proteins; Enzyme catalyzed polymerization and precipitation of aromatic compounds from wastewater.


Still not finding the right product?

Explore all of our products under Peroxidase from horseradish


pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Resp. Sens. 1

Storage Class

13 - Non Combustible Solids

wgk

WGK 1

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library