Skip to Content
Merck
All Photos(1)

Documents

P5521

Sigma-Aldrich

Phosphatase, Alkaline from bovine intestinal mucosa

≥2,000 DEA units/mg protein

Synonym(s):

Alkaline phosphatase, Orthophosphoric-monoester phosphohydrolase (alkaline optimum)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

form

aqueous solution

specific activity

≥2,000 DEA units/mg protein

mol wt

dimer ~160 kDa

concentration

≥1.0 mg/mL

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

The enzyme is a glycoprotein containing approximately 12% carbohydrate (6% hexoses and 6% other neutral sugars). Each molecule of alkaline phosphatase contains four zinc atoms and four disulfide bridges. Maximal activity with alkaline phosphatase is achieved in the presence of magnesium. It catalyzes the hydrolysis of phosphate monoesters such as p-nitrophenyl phosphate, phenyl phosphate, phenolphthalein phosphate, α-glycerol phosphate, β-glycerol phosphate, 2-phosphorylglycerate, triosephosphate, glucose-6-phosphate, glucose 1-phosphate, fructose 1-phosphate, fructose 6-phosphate, adenosine 5-phosphate adenosine 3-phosphate, phosphoenolpyruvate, and β-nicotinamide adenine dinucleotide phosphate. Arsenate, cysteine, iodine, inorganic phosphate, pyrophosphate, diisopropyl phosphate, triphenylphosphate, diisopropyl fluorophosphate, and L-phenylalanine are some of the strong inhibitors of alkaline phosphatase.

Application

Alkaline phosphatase can be used to dephosphorylate casein and other proteins. Alkaline phosphatase may be also be used to dephosphorylate the 5′-termini of DNA or RNA to prevent self-ligation. DNA or RNA can also be tagged with radiolabeled phosphate (via T4 polynucleotide kinase) after dephosphorylation with alkaline phosphatase..
Alkaline phosphatase is used for conjugation to antibodies and other proteins for ELISA, Western blotting, and histochemical detection. It is routinely used to dephosphorylate proteins, such as casein, and nucleic acids. It may be used for protein labeling when high sensitivity is required. Alkaline phosphatase may be also be used to dephosphorylate the 5′-termini of DNA or RNA to prevent self-ligation. DNA or RNA can also be tagged with radiolabeled phosphate (via T4 polynucleotide kinase) after dephosphorylation with alkaline phosphatase. Product P5521 has been used to study GTP hydrolysis .
High specific activity grade recommended for antibody and protein conjugation.
The enzyme from Sigma has been used in the assay of phosphatidylinositol 4-phosphate and phosphatidylinositol 4,5-bisphosphate. It has also been used for alkaline phosphatase assays using a Nafion-modified electrode.

Biochem/physiol Actions

Alkaline phosphatase, from bovine intestinal mucosa, is most stable in the pH range 7.5-9.5. The enzyme has a broad specificity for phosphate esters of alcohols, amines, pyrophosphate, and phenols and it requires zinc, and magnesium or calcium divalent ions for activity.
The enzyme has a broad specificity for phosphate esters of alcohols, amines, pyrophosphate, and phenols. It is routinely used to dephosphorylate proteins and nucleic acids.

Physical properties

Bovine intestinal alkaline phosphatase is a dimeric, membrane-derived glycoprotein. At least three isoforms exist, which typically possess two N-linked and one or more O-linked glycans per monomer.
Bovine intestinal alkaline phosphatase is a dimeric, membrane-derived glycoprotein. At least three isoforms exist, which typically possess two N-linked and one or more O-linked glycans per monomer.2 The enzyme requires zinc, and magnesium or calcium divalent ions for activity.

Unit Definition

One DEA unit will hydrolyze 1 μmole of 4-nitrophenyl phosphate per minute at pH 9.8 at 37 °C. (One glycine unit is equivalent to ~3 DEA units)

Physical form

solution in 3.2 M ammonium sulfate, 1 mM MgCl2 and 0.1 mM ZnCl2, pH 7.0

Preparation Note

Affinity purified
This product is prepared from bovine intestinal mucosa. Dilutions should be made in 10 mM Tris-HCl, pH 8.0, 1-5 mM magnesium chloride, 0.1-0.2 mM zinc chloride, and 50% glycerol and stored at 2-8 °C.

Analysis Note

Package sizes are based on DEA units
Protein determined by biuret.

Legal Information

Nafion is a trademark of The Chemours Company FC, LLC

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B A Ponder et al.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society, 31(7), 911-919 (1983-07-01)
The immunohistological demonstration of H2 antigens in cryostat sections of a wide variety of mouse tissues is reported. The purpose in developing the method was to use H2 antigens as cellular markers in studies of mouse chimeras. Monoclonal anti-H2 antibodies
P T Hawkins et al.
The Biochemical journal, 218(3), 785-793 (1984-03-15)
We have developed methods that yield estimates of the 32P content of each of the individual phosphate groups of phosphatidylinositol 4-phosphate and phosphatidylinositol 4,5-bisphosphate, thus extending the information available from studies of the labelling of these lipids in intact cells
Determination of alkaline phosphatase using a Nafion?-modified electrode.
Le Gal La Salle A, et al.
Journal of Electroanalytical Chemistry, 379(1), 281-291 (1994)
Alkaline phosphatase (EC 3.1.3.1).
M J Maunders
Methods in molecular biology (Clifton, N.J.), 16, 331-341 (1993-01-01)
Xinlin Du et al.
Proceedings of the National Academy of Sciences of the United States of America, 101(24), 8858-8863 (2004-06-05)
A remote labeling method has been developed to determine (18)O kinetic isotope effects (KIEs) in Ras-catalyzed GTP hydrolysis. Substrate mixtures consist of (13)C-depleted GTP and [(18)O,(13)C]GTP that contains (18)O at phosphoryl positions of mechanistic interest and (13)C at all carbon

Articles

Instructions for working with enzymes supplied as ammonium sulfate suspensions

Protocols

Enzymatic Assay of Alkaline Phosphatase, Diethanolamine Assay (EC 3. 1. 3. 1)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service