Skip to Content
Merck
All Photos(2)

Key Documents

N8630

Sigma-Aldrich

Nuclease P1 from Penicillium citrinum

lyophilized powder, ≥200 units/mg protein (E1%/280, 3′-5′-Phosphodiesterase)

Synonym(s):

3′-Phosphohydrolase, Nuclease 5′-Nucleotidehydrolase, Endonuclease P1

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

biological source

Penicillium citrinum

Quality Level

form

lyophilized powder

specific activity

≥200 units/mg protein (E1%/280, 3′-5′-Phosphodiesterase)

secondary activity

≥1,000 units/mg protein 3′-nucleotidase

mol wt

42-50 kDa

packaging

vial of ≥250 units (using RNA substrate)

technique(s)

DNA extraction: suitable
DNA purification: suitable

suitability

suitable for molecular biology

application(s)

cell analysis

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Nuclease P1 is one of the most commonly known single-strand specific nucleases in molecular biology; Nuclease P1 is a single stranded specific endoduclease (of ssDNA or ssRNA). Nuclease P1 can also cleave single-stranded regions in double-stranded nucleic acids.
Nuclease P1 from Penicillium citrinum is a zinc-dependent endonuclease that exhibits increased activity in the presence of low concentrations of urea. Nuclease P1 selective activity has found useful applications in studies on nucleic acid structure.

Application

  • Nuclease P1 cleave of single stranded DNA or RNA to 5′ mononucleotides
  • Nuclease P1 supports DNA damage and modification research
  • Nucleic acids base composition and structural analysis can be done by Nuclease P1
  • Nuclease P1 has historically been used for the industrial production of 5′-mononucleotides from yeast RNA.
  • Removal of nucleic acids through protein purification can be done by Nuclease P1
  • Nuclease P1 is a key reagent for the development of methods for studies involving t-RNA dependent amino acid biosynthesis and t-RNA dependent trans-amidation
  • Nuclease P1 from Penicillium citrinum has been used in a study to assess crystal structures using ammonium sulphate or polyethylene glycol 4000 as a precipitating agent.
  • Nuclease P1 was used in a study to investigate a method for the direct sequence analysis 20-25 nucleotides from the terinini of 5′ or 3′ end group labeled RNA.
  • Nuclease P1 is used to improve the sensitivity of a 32P-labeling method for the detection of DNA adducts.
The enzyme has an optimal temperature of approximately 70°C, but for a long incubation, a temperature below 60°C is more suitable. It is stable in the pH range of 5 - 8.
The enzyme has an optimal temperature of approximately 70 °C, but for a long incubation, a temperature below 60 °C is more suitable. It is stable in the pH range of 5 - 8.

Biochem/physiol Actions

Catalyzes the nonspecific endonucleolytic cleavage of single stranded DNA and RNA to yield nucleoside 5′-phosphates and 5′-phosphooligonucleotides. It does not appreciably degrade double-stranded nucleic acids, especially in the presence of more than 400 mM sodium chloride at pH 6.0.

Features and Benefits

Our highly active Nuclease P1 is tested for its 3′- 5′ - Phosphodiesterase Activity and 3′- Nucleotidase Activity and is the most active Nuclease P1 in the market

Physical properties

A zinc dependent glycoprotein consisting of 270 amino acid residues. Molecular mass: 42-50 kDa.

Unit Definition

3′-5′-Phosphodiesterase: One unit will liberate 1.0 μmole of acid soluble nucleotides from RNA per min at pH 5.3 at 37 °C.
3′-Nucleotidase: One unit will hydrolyze 1.0 μmole of orthophosphate from 3′-AMP per min at pH 7.2 at 37 °C.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fahimeh Salehi et al.
Scientific reports, 8(1), 13902-13902 (2018-09-19)
DNA targeting anticancer agents have been very successful in clinic, especially, when used in combinatorial therapy. But unfortunately, they often exhibit high levels of toxicity towards normal cells. Hence, much effort has been put into finding agents with more selectivity
Xiaohuan Jin et al.
Nucleic acids research, 47(2), 883-898 (2018-12-07)
Modified nucleosides on tRNA are critical for decoding processes and protein translation. tRNAs can be modified through 1-methylguanosine (m1G) on position 37; a function mediated by Trm5 homologs. We show that AtTRM5a (At3g56120) is a Trm5 ortholog in Arabidopsis thaliana.
K Shanmugha Rajan et al.
Nucleic acids research, 47(14), 7633-7647 (2019-05-31)
The parasite Trypanosoma brucei, the causative agent of sleeping sickness, cycles between an insect and a mammalian host. Here, we investigated the presence of pseudouridines (Ψs) on the spliceosomal small nuclear RNAs (snRNAs), which may enable growth at the very
A Lahm et al.
Journal of molecular biology, 215(2), 207-210 (1990-09-20)
P1 nuclease, a zinc-dependent single-strand specific endonuclease from Penicillium citrinum, has been crystallized in three different space groups using either ammonium sulphate or polyethylene glycol 4000 as the precipitating agent. The crystals diffract to between 3 A and 2.2 A.
Enhancement of nuclease P1 activity in low concentration of denaturants
Gangadhara, K. and P. Gangadhara
Enzyme and Microbial Technology, 43, 7-7 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service