Skip to Content
Merck
All Photos(4)

Documents

P7640

Sigma-Aldrich

Phosphatase, Alkaline from bovine intestinal mucosa

lyophilized powder, ≥10 DEA units/mg solid

Synonym(s):

Alkaline phosphatase, Orthophosphoric-monoester phosphohydrolase (alkaline optimum)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
eCl@ss:
42010105
NACRES:
NA.54

biological source

bovine intestinal mucosa

form

lyophilized powder

specific activity

≥10 DEA units/mg solid

mol wt

dimer ~160 kDa

shipped in

wet ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

Alkaline phosphatase can be used to dephosphorylate casein and other proteins. Alkaline phosphatase may be also be used to dephosphorylate the 5′-termini of DNA or RNA to prevent self-ligation. DNA or RNA can also be tagged with radiolabeled phosphate (via T4 polynucleotide kinase) after dephosphorylation with alkaline phosphatase..
Alkaline phosphatase is used for conjugation to antibodies and other proteins for ELISA, Western blotting, and histochemical detection. It is routinely used to dephosphorylate proteins and nucleic acids. It may be used for protein labeling when high sensitivity is required. Alkaline phosphatase may be also be used to dephosphorylate the 5′-termini of DNA or RNA to prevent self-ligation. DNA or RNA can also be tagged with radiolabeled phosphate (via T4 polynucleotide kinase) after dephosphorylation with alkaline phosphatase.. Product P7640 has been used to study the monoclonal alkaline phosphatase-anti-alkaline phosphatase (APAAP) complex.
High specific activity grade recommended for antibody and protein conjugation.

Biochem/physiol Actions

Alkaline phosphatase, from bovine intestinal mucosa, is most stable in the pH range 7.5-9.5. The enzyme has a broad specificity for phosphate esters of alcohols, amines, pyrophosphate, and phenols and it requires zinc, and magnesium or calcium divalent ions for activity.
The enzyme has a broad specificity for phosphate esters of alcohols, amines, pyrophosphate, and phenols. It is routinely used to dephosphorylate proteins and nucleic acids.

Physical properties

Bovine intestinal alkaline phosphatase is a dimeric, membrane-derived glycoprotein. At least three isoforms exist, which typically possess two N-linked and one or more O-linked glycans per monomer.
Bovine intestinal alkaline phosphatase is a dimeric, membrane-derived glycoprotein. At least three isoforms exist, which typically possess two N-linked and one or more O-linked glycans per monomer.2 The enzyme requires zinc, and magnesium or calcium divalent ions for activity.

Unit Definition

One DEA unit will hydrolyze 1 μmole of 4-nitrophenyl phosphate per minute at pH 9.8 at 37 °C. (One glycine unit is equivalent to ~3 DEA units)

Analysis Note

Package sizes are based on DEA units

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mamoni Dash et al.
Carbohydrate polymers, 182, 254-264 (2017-12-28)
Polyelectrolyte complexes (PEC) of chitosan and ulvan were fabricated to study alkaline phosphatase (ALP) mediated formation of apatitic minerals. Scaffolds of the PEC were subjected to ALP and successful mineral formation was studied using SEM, Raman and XRD techniques. Investigation
Simultaneous flow-injection determination of fluoride, monofluorophosphate and orthophosphate ions using alkaline phosphatase immobilized on a cellulose nitrate membrane and an open-circulation approach.
Tzanavaras PD and Themelis DG
Analytica Chimica Acta, 467 (1-2), 83-89 (2002)
A Hohmann et al.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society, 36(2), 137-143 (1988-02-01)
A mouse monoclonal antibody, FMC55 (an IgG1), to alkaline phosphatase was prepared and evaluated in immunostaining. Clones producing antibody to alkaline phosphatase were selected using a micro-ELISA which identified antibodies forming active soluble complexes (APAAP) with the enzyme. Conditions that
Stefanie Riedel et al.
Journal of functional biomaterials, 12(4) (2021-10-27)
Biological hydrogels are highly promising materials for bone tissue engineering (BTE) due to their high biocompatibility and biomimetic characteristics. However, for advanced and customized BTE, precise tools for material stabilization and tuning material properties are desired while optimal mineralisation must
Some properties of alkaline phosphatase of cow's milk and calf intestinal mucosa.
R K MORTON
The Biochemical journal, 60(4), 573-582 (1955-08-01)

Protocols

Enzymatic Assay of Alkaline Phosphatase, Diethanolamine Assay (EC 3. 1. 3. 1)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service